Recommended Products
biological source
fish brain
packaging
tube of 5 μg 10072802-DNA-5UG
pkg of vial of cells 10072802-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Squamous
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
Cell Line Origin
Common carp brain cell line, koi herpesvirus isolation
Cell Line Description
The CCB cell line was developed from Common carp (Cyprinus carpio carpio) brain.
Packaging
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Culture Medium
EMEM (EBSS) + 2 mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Fetal Bovine Serum (FBS). 10 mM HEPES can be included if a CO2 incubator is not available.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:6, i.e., seeding at 2 x 104 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 25 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Irene Cano et al.
International journal of molecular sciences, 21(22) (2020-11-15)
Seroconversion and the mucosal lysozyme G (lysG), complement 3 (c3), and immunoglobulins M (IgMsec) and Z2 (IgZ2) were measured for up to 900 degree days (DD) in skin swabs from common carp exposed to koi herpesvirus (KHV or CyHV-3) at
Irene Cano et al.
Animals : an open access journal from MDPI, 11(2) (2021-02-13)
Fluorescence real-time LAMP assays were designed for the orf43 gene of CyHV-3 European genotype and the p4a gene of the CEV genogroup I. A third LAMP assay to detect the ef1a gene of the host common carp was designed as
Agnieszka Troszok et al.
Journal of fish diseases, 41(11), 1709-1718 (2018-08-26)
Cyprinid herpesvirus 3 (CyHV-3), also known as koi herpesvirus (KHV), is an aetiological agent of a virulent and lethal disease in common and koi carp. In this study, we examined in vitro the anti-CyHV-3 activity of acyclovir (ACV), nucleoside analogue commonly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service