Skip to Content
Merck

Skip To

10070

α-Amylase from Bacillus sp.

greener alternative

Greener Alternative

powder, yellow-brown, ~50 U/mg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-565-6
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
~50 U/mg
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

specific activity

~50 U/mg

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

yellow-brown

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch hydrolysis research. For more information see the article in biofiles.

Other Notes

Applications in (selective) hydrolysis/condensations of glycosidic bonds.
One unit corresponds to the amount of enzyme which liberates 1 μmole maltose per minute at pH 6.9 at 25 °C (starch CAS 9005-25-8).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10069A6814A1031
description

powder, yellow-brown, ~50 U/mg

description

powder, yellow-brown, ~380 U/mg

description

powder, ≥400 units/mg protein (Lowry)

description

Type XIII-A, lyophilized powder, 300-1,500 units/mg protein

specific activity

~50 U/mg

specific activity

~380 U/mg

specific activity

≥400 units/mg protein (Lowry)

specific activity

300-1,500 units/mg protein

form

powder

form

powder

form

powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

color

yellow-brown

color

yellow-brown

color

-

color

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

greener alternative category

Enabling,

greener alternative category

, Enabling

greener alternative category

-

greener alternative category

Enabling,


Still not finding the right product?

Explore all of our products under α-Amylase from Bacillus sp.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Welche Uniprot Nummer hat dieses Protein?

    1 answer
    1. Unfortunately, this product does not have a UniProt number.

      Helpful?

Reviews

No rating value

Active Filters