Skip to Content
Merck

07371

Novel Juice, DNA stain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

storage temp.

−20°C

General description

Novel Juice is a non-mutagenic fluorescent reagent that produces instant visualization of DNA bands upon Blue Light or UV illumination of agarose gels. Supplied in GeneDireX′s 6X DNA Loading Buffer, Novel Juice is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. Novel Juice is the most sensitive stain available for detecting the double-stranded DNA (dsDNA). It contains three tracking dyes (Bromophenol Blue, Xylene Cyanol FF, and Orange G) for visually tracking the DNA migration during the electrophoresis process. It is ideal for the environment requiring a safe, non-hazardous alternative to Ethidium Bromide.


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library