Skip to Content
Merck

07188

Lithium mupirocin

≥95.0% (HPLC)

Synonym(s):

9-((E)-4-{(2S,3R,4R,5S)-3,4-Dihydroxy-5-[(2S,3S)-3-((1S,2S)-2-hydroxy-1-methyl-propyl)oxiranylmethyl]tetrahydro-pyran-2-yl}-3-methyl-but-2-enoyloxy)nonanoic acid lithium salt, Pseudomonic acid A lithium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H43LiO9
CAS Number:
Molecular Weight:
506.56
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (HPLC)

form

powder

color

white to off-white

antibiotic activity spectrum

Gram-positive bacteria

mode of action

protein synthesis | interferes

SMILES string

[Li+].C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(C)=C\C(=O)OCCCCCCCCC([O-])=O)[C@H](O)[C@@H]2O

InChI

1S/C26H44O9.Li/c1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27;/h13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29);/q;+1/p-1/b16-13+;/t17-,18-,19-,20-,21-,24+,25-,26-;/m0./s1

InChI key

XFIKMPRBSORKQP-JATHGWPISA-M

Application

Lithium mupirocin as been used to study mupirocin resistance in staphylococcus aureus and in mycoplasma susceptibility studies .

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place. Storage class (TRGS 510): Non Combustible Solids


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library