Skip to Content
Merck

07077

N-(p-Coumaroyl)-L-homoserine lactone

≥94% (HPLC)

Synonym(s):

p-Coumaroyl (S)-(−)α-amino-γ-butyrolactone, pC-HSL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H13NO4
Molecular Weight:
247.25
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥94% (HPLC)

form

powder

optical activity

[α]/D -23.0±3.0°, c = 1% in ethanol

color

white to off-white

mp

201.2 °C

application(s)

cell analysis

storage temp.

2-8°C

InChI

1S/C13H13NO4/c15-10-4-1-9(2-5-10)3-6-12(16)14-11-7-8-18-13(11)17/h1-6,11,15H,7-8H2,(H,14,16)/b6-3+/t11-/m0/s1

InChI key

CCIXZFJYFQJTGK-GQOHGMTASA-N

Application

N-(p-Coumaroyl)-L-homoserine lactone, or pC-HSL, can be used to study microbiology, quorum sensing and signaling compounds. pC-HSL has been used to study the activity of the Rhodopseudomonas palustris p-coumaroyl-homoserine lactone-responsive transcription factor RpaR.

Biochem/physiol Actions

New type of bacterial, quorum sensing signal compound. Quorum sensing mechanism has been investigated in Rhodopseudomonas palustris, but the signal compound has been discovered in other bacteria such as Bradyrhizobium sp. or Silicibacter pomeroyi.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library