Skip to Content
Merck
All Photos(1)

Key Documents

06627

Sigma-Aldrich

2-Aminoacridone

BioReagent, suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

2-Amino-9(10H)-acridinone, 2-amino-10H-acridin-9-one, AMAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O
CAS Number:
Molecular Weight:
210.23
Beilstein:
172520
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥98.0% (HPLC)

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 420 nm; λem 542 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

Nc1ccc2Nc3ccccc3C(=O)c2c1

InChI

1S/C13H10N2O/c14-8-5-6-12-10(7-8)13(16)9-3-1-2-4-11(9)15-12/h1-7H,14H2,(H,15,16)

InChI key

PIGCSKVALLVWKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Aminoacridone is a highly fluorescent aromatic, which contains a primary amine group that reacts with an aldehyde group at the reducing end of a carbohydrate and is reduced to a stable amine derivative by sodium borohydride (NaBH4). Picomolar levels of glycan compounds can be detected using this fluorophore. The resulting derivatzed compounds can be separated by reverse-phase HPLC and detected by positive-ion electrospray MS . An intense fluorescent, hydrophobic probe that is stable over a wide pH range is useful in the derivatization of glycans to allow for the analysis of complex oligosaccharides using micellar electrokinetic capillary chromatography and reverse- and normal-phase chromatography coupled with mass spectroscopy to determine relative concentrations and structural identity of individual oligosaccharides . The λ excitation and λ emission are 425 nm and 532 nm, respectively.
Fluorescent label for glycans and saccharides.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoji Oonuki et al.
Analytical biochemistry, 343(2), 212-222 (2005-07-12)
Various combinations of fluorescent dyes, polyacrylamide gels, and electrophoresis buffers were tested by fluorophore-assisted carbohydrate electrophoresis (FACE) for the purpose of analyzing sulfated and nonsulfated glycosaminoglycan (GAG) oligosaccharides in which disaccharides and low-molecular weight oligosaccharides were included. A nonionic fluorescent
Vojtech Franc et al.
Journal of the American Society for Mass Spectrometry, 29(6), 1099-1110 (2018-03-14)
The human complement hetero-trimeric C8αβγ (C8) protein assembly (~ 150 kDa) is an important component of the membrane attack complex (MAC). C8 initiates membrane penetration and coordinates MAC pore formation. Here, we charted in detail the structural micro-heterogeneity within C8, purified from
Martin Giera et al.
The Analyst, 136(13), 2763-2769 (2011-05-26)
Malondialdehyde (MDA) has become a well-established biomarker for oxidative stress. The most commonly used way to determine urinary MDA levels is the thiobarbituric acid (TBA) assay, which suffers from several drawbacks. In this manuscript, we describe a novel derivatization strategy
Alicia M Hitchcock et al.
Electrophoresis, 29(22), 4538-4548 (2008-11-28)
This work describes improved workup and instrumental conditions to enable robust, sensitive glycosaminoglycan (GAG) disaccharide analysis from complex biological samples. In the process of applying CE with LIF to GAG disaccharide analysis in biological samples, we have made improvements to
Yong Zhang et al.
Electrophoresis, 28(3), 414-421 (2007-03-16)
To improve the separation of monosulfate glycosaminoglycan disaccharide isomers by microchip electrophoresis, we found that addition of 1,4-dioxane (DO) dramatically improved analyte resolution, probably due to solvation effects. Methylcellulose (MC) was tested for the ability to suppress EOF and analyte

Articles

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service