Skip to Content
MilliporeSigma

Skip To

02071

Catalase from Corynebacterium glutamicum

solution, deep brown, ≥500000 U/mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Check Cart for Availability
$78.80
5 mL
Check Cart for Availability
$295.00

About This Item

CAS Number:
NACRES:
NA.54
UNSPSC Code:
12352204
MDL number:
EC Number:
eCl@ss:
32160410
Form:
solution

$78.80


Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0041

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Corynebacterium glutamicum)

Quality Segment

form

solution

concentration

≥500000 U/mL

color

deep brown

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O(CC)C(=O)c1ccc(cc1)O

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

General description

Catalase is ubiquitously found in animals, plants, fungi, and humans. It is made up of looped polypeptide chains that are arranged as tetramers or dimers and a core, that contains iron or manganese ions. Catalase can be differentiated into two types such as heme-containing (type I), which possesses an iron-porphyrin cofactor and a non-heme-containing (type II), which possesses a dinuclear manganese active site.

Application

Catalase from Corynebacterium glutamicum has been used to identify and characterize purified bacterial peptide (PPBP) produced from Bacillus thuringiensis. It has also been used to study its inhibition by nitric oxide.

Biochem/physiol Actions

Catalase is an antioxidant enzyme that plays a role in alleviating oxidative stress by the removal of hydrogen peroxide to produce oxygen and water. Lower levels of catalase lead to several age-related disorders including hypertension, anemia, vitiligo, Parkinson′s disease, Alzheimer′s disease, schizophrenia, bipolar disorders, diabetes mellitus, and cancer.

Physical form

contains ~30% glycerol, 10% ethanol

Other Notes

1 U corresponds to the amount of enzyme which decomposes 1 μmol H2O2 per minute at pH 7.0 and 25°C
The purity of this product is not determined. It is tested and sold by activity rather than mass.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
60634C3155C30
form

solution

form

solution

form

aqueous solution

form

aqueous suspension

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

deep brown

color

dark brown

color

-

color

-

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

biological source

bacterial (Corynebacterium glutamicum)

biological source

bacterial (Micrococcus lysodeikticus)

biological source

bovine liver

biological source

bovine liver


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

105.8 °F - closed cup

flash_point_c

41 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the concentration and purity of the catalase?

    1 answer
    1. The purity of this product is not determined. It is tested and sold by activity rather than mass. The lot specific activity is reported in units per mL and can be found in the Certificate of Analysis. Please see the link below for sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/PDF736758.pdf

      Helpful?

Reviews

No rating value

Active Filters