Skip to Content
Merck

01383

UV-VIS detectable IEF Marker solution

pI 3.9, 1 mg/mL in 10 mM NaOH, 0.45 μm filtered

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

0.45 μm filtered

concentration

1 mg/mL in 10 mM NaOH

storage temp.

2-8°C

Application

suitable as IEF-Marker for HPCE and for IEF-gels


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library