Skip to Content
Merck
All Photos(5)

Key Documents

S9390

Sigma-Aldrich

Sodium phosphate dibasic heptahydrate

ACS reagent, 98.0-102.0%

Synonym(s):

Disodium hydrogen phosphate heptahydrate, Disodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder

grade

ACS reagent

Quality Level

vapor density

4.9 (vs air)

Assay

98.0-102.0%

form

powder

impurities

≤0.005% Insoluble matter

pH

8.7-9.3 (25 °C, 5% in solution)

density

1.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals: ≤0.001% (by ICP)

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.7H2O/c;;1-5(2,3)4;;;;;;;/h;;(H3,1,2,3,4);7*1H2/q2*+1;;;;;;;;/p-2

InChI key

PYLIXCKOHOHGKQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate dibasic heptahydrate is a hydrated form of disodium phosphate (DSP) or disodium hydrogen orthophosphate. DSP is widely used as a pH regulator and saline laxative.

Application

A mixture of sodium phosphate dibasic heptahydrate, potassium phosphate monobasic, and NaCl in deionized water is used for the preparation of phosphate-buffered saline (PBS, pH 7.4). PBS is used as a buffer solution in biological research to keep the pH constant.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina Petrova et al.
Scientific reports, 9(1), 8745-8745 (2019-06-21)
Circulating cancer markers are metabolic products found in body fluids of cancer patients, which are specific for a certain type of malignant tumors. Cancer marker detection plays a key role in cancer diagnosis, treatment, and disease monitoring. The growing need
Christian M Schürch et al.
Cell, 182(5), 1341-1359 (2020-08-09)
Antitumoral immunity requires organized, spatially nuanced interactions between components of the immune tumor microenvironment (iTME). Understanding this coordinated behavior in effective versus ineffective tumor control will advance immunotherapies. We re-engineered co-detection by indexing (CODEX) for paraffin-embedded tissue microarrays, enabling simultaneous
Francesca Vallese et al.
Nature communications, 11(1), 6069-6069 (2020-11-29)
Membrane contact sites between virtually any known organelle have been documented and, in the last decades, their study received momentum due to their importance for fundamental activities of the cell and for the subtle comprehension of many human diseases. The
Sebastian Baumann et al.
Science advances, 6(11), eaaz1588-eaaz1588 (2020-03-24)
Through the asymmetric distribution of messenger RNAs (mRNAs), cells spatially regulate gene expression to create cytoplasmic domains with specialized functions. In neurons, mRNA localization is required for essential processes such as cell polarization, migration, and synaptic plasticity underlying long-term memory
Ziyi Yin et al.
Autophagy, 16(5), 900-916 (2019-07-18)
Cells are faced with various stresses during their growth and development, and autophagy is a degradative process in which cells can break down their own components to recycle macromolecules and provide energy under these stresses. For pathogenic fungi that utilize

Articles

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service