Skip to Content
Merck
All Photos(2)

Key Documents

I9030

Sigma-Aldrich

2-Propanol

≥99.0%, meets USP testing specifications, USP/NF

Synonym(s):

2-Propanol, isopropanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

Isopropyl alcohol, meets USP testing specifications

Agency

USP/NF
meets USP testing specifications

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

dilution

(for analytical testing)

impurities

≤0.5% (water)

refractive index

n20/D 1.377 (lit.)

pH

7.0 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl alcohol (2-Propanol, i-PrOH), a renewable secondary alcohol, is water soluble, colorless, volatile liquid with a sweet odor. It is a potential substitute to petroleum-derived gasoline. The preparation of methyl isobutyl ketone (MIBK) using i-PrOH as the precursor has been reported with yield as high as 25%. The solubility of benzoic acid has been investigated in i-PrOH by gravimetric method in the temperature range, 277-346K. Its photocatalytic degradation has been examined by various methods. This product is a high quality pharmacopoeia product that meets the testing specifications of USP (United States Pharmacopoeia). It can be employed in research and pilot studies.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Photocatalytic degradation of 2-propanol and phenol using Au loaded MnWO4 nanorod under visible light irradiation.
Chakraborty AK, et al.
Journal of Cluster Science, 23(2), 437-448 (2012)
Photocatalytic degradation of 2-propanol under irradiation of visible light by nanocrystalline titanium (IV) oxide modified with rhodium ion using adsorption method.
Kitano S, et al.
Chemistry Letters (Jpn), 39(6), 627-629 (2010)
Influence of activated carbon in TiO2 and ZnO mediated photo-assisted degradation of 2-propanol in gas-solid regime.
Matos J, et al.
Applied Catalysis. B, Environmental, 99(1), 170-180 (2010)
Solubility of benzoic acid in acetone, 2-propanol, acetic acid and cyclohexane: Experimental measurement and thermodynamic modeling.
Long B, et al.
Fluid Phase Equilibria, 297(1), 113-120 (2010)
2-propanol oxidation in a pressurized jet-stirred reactor (JSR) and combustion bomb: experimental and detailed kinetic modeling study.
Togbe C, et al.
Energy and Fuels, 25(2), 676-683 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service