Skip to Content
Merck
All Photos(3)

Key Documents

D221104

Sigma-Aldrich

Dodecane

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
Molecular Weight:
170.33
Beilstein:
1697175
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
technique(s):
microbiological culture: suitable
bp:
215-217 °C (lit.)
vapor pressure:
1 mmHg ( 47.8 °C)

grade

reagent

Quality Level

vapor density

5.96 (vs air)

vapor pressure

1 mmHg ( 47.8 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

401 °F

expl. lim.

8 %

technique(s)

microbiological culture: suitable

refractive index

n20/D 1.421 (lit.)

bp

215-217 °C (lit.)

mp

−9.6 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC

InChI

1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

SNRUBQQJIBEYMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dodecane is a long-chain linear alkane that is commonly used as a solvent and the major constituent of kerosene. Also, it is employed as an organic phase in solvent extraction.

Application

Dodecane can be used as a solvent in the:
  • Extraction of hydrocarbons from Botryococcus braunii.
  • Pd/C catalyzed deoxygenation of stearic acid to synthesize biodiesel.
  • Oxidativede sulfurization of dibenzothiophenes.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F - Pensky-Martens closed cup

Flash Point(C)

70 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gina M Geiselman et al.
Microbial cell factories, 19(1), 24-24 (2020-02-07)
Rhodosporidium toruloides has emerged as a promising host for the production of bioproducts from lignocellulose, in part due to its ability to grow on lignocellulosic feedstocks, tolerate growth inhibitors, and co-utilize sugars and lignin-derived monomers. Ent-kaurene derivatives have a diverse
X-L Cao et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(6), 791-798 (2011-05-31)
A total of 154 food composite samples from the 2008 total diet study in Quebec City were analysed for bisphenol A (BPA), and BPA was detected in less than half (36%, or 55 samples) of the samples tested. High concentrations
Raquel Silva et al.
Advances in experimental medicine and biology, 733, 155-164 (2011-11-22)
In this work, we report a novel approach using proteinaceous microspheres of bovine serum albumin (BSA), human serum albumin (HSA) and silk fibroin (SF) containing different organic solvents, namely n-dodecane, mineral oil and vegetable oil, to reduce the activity of
Masoumeh Karvar et al.
Langmuir : the ACS journal of surfaces and colloids, 27(17), 10386-10391 (2011-07-07)
Surfactants such as Aerosol OT (AOT) are commonly used to stabilize and electrically charge nonpolar colloids in devices such as electronic ink displays. The electrical behavior of such devices is strongly influenced by the presence of charged inverse micelles, formed
Dorinde M M Kleinegris et al.
Enzyme and microbial technology, 48(3), 253-259 (2011-11-25)
During the in situ extraction of β-carotene from Dunaliella salina, the causal relationship between carotenoid extraction and cell death indicated that cell growth and cell death should be at equilibrium for a continuous in situ extraction process. In a flat-panel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service