Skip to Content
Merck
All Photos(7)

Documents

791776

Sigma-Aldrich

Potassium carbonate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.005% silica
≤0.01% insolubles

pH

11-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001%
sulfur compounds (as SO42-): ≤0.004%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
Mg: ≤0.002%
Na: ≤0.02%
heavy metals (as Pb): ≤5 ppm

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium carbonate is an inorganic base. It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene. Addition of amino acids (glycine, sarcosine and proline) to K2CO3 (solvent) promotes the absorption of CO2.

Application

Potassium carbonate has been used in the preparation of disulfonated bis[4-(3-aminophenoxy)phenyl]sulfone, a disulfonated diamine monomer.

It may be used:
  • As a reagent in the synthesis of polysubstituted iodobenzene derivatives.
  • As a base for the synthesis of high molecular weight homopolymers and copolymers derived from various bisphenols.
  • As a base for the Suzuki coupling of aryl halides with aryl boronic acids.
  • In the Heck reaction of styrene and bromobenzene.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam F Nugraha et al.
Polymers, 12(7) (2020-07-24)
Several methods to synthesize poly(phenylene) block copolymers through the nickel coupling reaction were attempted to reduce the use of expensive nickel catalysts in polymerization. The model reaction for poly(phenylene) having different types of dichlorobenzene derivative monomers illustrated the potential use
Yusuke Yagi et al.
Organic & biomolecular chemistry, 13(4), 1113-1121 (2014-11-26)
Fluorine-18 labeled radiotracers, such as [(18)F]fluorodeoxyglucose, can be used as practical diagnostic agents in positron emission tomography (PET). Furthermore, the properties of pharmaceuticals can be enhanced significantly by the introduction of fluorine groups into their original structures, and significant progress
Brian Capozzi et al.
Journal of the American Chemical Society, 136(29), 10486-10492 (2014-07-09)
We have measured the single-molecule conductance of a family of oligothiophenes comprising 1-6 thiophene moieties terminated with methyl-sulfide linkers using the scanning tunneling microscope-based break-junction technique. We find an anomalous behavior: the peak of the conductance histogram distribution does not
K M Frankowski et al.
Journal of dairy science, 97(9), 5356-5370 (2014-07-16)
Many food companies are trying to limit the amount of sodium in their products. Permeate, the liquid remaining after whey or milk is ultrafiltered, has been suggested as a salt substitute. The objective of this study was to determine the
Ellen Dirkx et al.
Journal of molecular and cellular cardiology, 76, 208-217 (2014-09-01)
In the insulin resistant heart, energy fuel selection shifts away from glucose utilization towards almost complete dependence on long-chain fatty acids (LCFA). This shift results in excessive cardiac lipid accumulation and eventually heart failure. Lipid-induced cardiomyopathy may be averted by

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service