Skip to Content
Merck
All Photos(1)

Key Documents

695068

Sigma-Aldrich

Hydrofluoric acid

ACS reagent, 48%

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

Assay

48%

form

liquid

density

1.15 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
phosphate (PO43-): ≤1 ppm
sulfate, sulfite (as SO42-): ≤5 ppm

cation traces

As: ≤0.05 ppm
Cu: ≤0.1 ppm
Fe: ≤1 ppm
heavy metals (as Pb): ≤0.5 ppm

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrofluoric acid is a solution of hydrogen fluoride (HF) in water. It is composed of hydrogen and fluoride. Hydrofluoric acid is a colorless and highly corrosive liquid. It is commonly used in the manufacture of hydrocarbons and polishing and conditioning glass. Additionally, it is used as a catalyst and in the synthesis of fluoride-containing compounds, and semiconductors.

Application

Hydrofluoric acid solution can be used as a reagent to synthesize:
  • Cyanohydrin alkyl ethers via chiral Bronsted acid-mediated hydrocyanation of vinyl ethers with silyl cyanides.
  • N, N-Dimethylaniline hydrochloride salts via Pd-catalyzed Buchwald-Hartwig amination of aryl triflates with dimethylamines.
It can also be used as a catalyst for the amidation of carboxylic acids with amines to synthesize amides.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrofluoric acid
DW Jr S, et al.
Elsevier's Integrated Review Biochemistry (Second Edition) null
Hydrofluoric acid burns
M H, et al.
Burns : Journal of the International Society For Burn Injuries, 30, 156-159 (2004)
Highly-ordered TiO2 nanotube arrays up to 220μm in length: use in water photoelectrolysis and dye-sensitized solar cells.
Shankar K, et al.
Nanotechnology, 18(6), 065707-065707 (2007)
Anatase TiO2 nanosheet: an ideal host structure for fast and efficient lithium insertion/extraction.
Chen JS and Lou XW.
Electrochemical Communications, 11(12), 2332-2335 (2009)
Francesca Monticelli et al.
Dental materials : official publication of the Academy of Dental Materials, 22(7), 602-609 (2005-11-18)
To verify the influence of different etching procedures of the post-surface on microtensile bond strength values between fiber posts and composite core materials. 60 DT Light Posts were divided into 10 subgroups using five different chemical surface treatments and two

Articles

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service