Skip to Content
Merck
All Photos(1)

Key Documents

62580

Sigma-Aldrich

Lithium perchlorate

purum p.a., ≥98.0% (calc. based on dry substance, T), powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiClO4
CAS Number:
Molecular Weight:
106.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥98.0% (calc. based on dry substance, T)

form

powder

reaction suitability

reagent type: oxidant

impurities

≤3% water

pH

6.0-9.5 (25 °C, 106.4 g/L)

mp

236 °C (lit.)

solubility

alcohol: soluble(lit.)
water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Li+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Li/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

MHCFAGZWMAWTNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium perchlorate (LiClO4) is a colorless lithium salt. Upon crystallization from its aqueous solution, it affords lithium perchlorate trihydrate (LiClO4.3H2O). It can be synthesized by reacting lithium chloride with perchloric acid.

Application

Lithium perchlorate (LiClO4) may be used as a catalyst in the synthesis of bis(indolyl)methanes and in the Baylis-Hillman coupling reaction of α,β-unsaturated carbonyl compounds with aldehydes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium perchlorate-accelerated Baylis-Hillman reactions.
Kawamura M and Kobayashi S.
Tetrahedron Letters, 40(8), 1539-1542 (1999)
Lithium perchlorate catalyzed reactions of indoles: An expeditious synthesis of bis (indolyl) methanes.
Yadav JS, et al.
Synthesis, 2001(5), 0783-0787 (2001)
Jun-Sik Seo et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(8), 1747-1756 (2002-08-09)
Vibrational characteristics of CD3CN solutions of LiClO4 and NaClO4 have been studied by means of infrared and Raman spectroscopy. Blue shifts of 22 and 11 cm(-1) of the v2 C[triple bond]N stretch are observed resulting from interaction of CD3CN with
Increased T(c) in electrolyte-gated cuprates.
Anoop Singh Dhoot et al.
Advanced materials (Deerfield Beach, Fla.), 22(23), 2529-2533 (2010-05-07)
Electrochemical enol ether/olefin cross-metathesis in a lithium perchlorate/nitromethane electrolyte solution.
Teppei Miura et al.
Angewandte Chemie (International ed. in English), 45(9), 1461-1463 (2006-01-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service