433314
Pyrophosphoric acid
technical grade
Synonym(s):
Diphosphoric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HO)2P(O)OP(O)(OH)2
CAS Number:
Molecular Weight:
177.98
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
grade
technical grade
Assay
≥94% (by NaOH, titration)
form
liquid
solid
waxy solid
technique(s)
ion chromatography: suitable
impurities
≥20% H3PO4
SMILES string
OP(O)(=O)OP(O)(O)=O
InChI
1S/H4O7P2/c1-8(2,3)7-9(4,5)6/h(H2,1,2,3)(H2,4,5,6)
InChI key
XPPKVPWEQAFLFU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Pyrophosphoric acid (diphosphoric acid) may be used in the following processes:
- As chemical activator in the preparation of activated carbon from leather waste.
- To modify the surface of calcium hydroxyapatite in order to improve the surface functionality.
- To modify the surface strontium aluminate nanoparticles potential applications in super-long time in vivo imaging.
- Synthesis of bis(diisopropylammonium) dihydrogendiphosphate trihydrate (DIPAP).
- Synthesis of novel 3-methoxybenzylammonium cation diphosphate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Persistent luminescent nanoparticles for super-long time in vivo and in situ imaging with repeatable excitation.
Sun M, et al.
Journal of Luminescence, 145, 838-842 (2014)
Surface modification of calcium hydroxyapatite with pyrophosphoric acid.
Tanaka H, et al.
Journal of Colloid and Interface Science, 269(2), 358-363 (2004)
Preparation and characterization of activated carbon from leather waste microwave-induced pyrophosphoric acid activation.
Kong J, et al.
Journal of Analytical and Applied Pyrolysis, 104, 710-713 (2013)
Structure of synthetic calcium hydroxyapatite particles modified with pyrophosphoric acid.
Tanaka H, et al.
Journal of Colloid and Interface Science, 283(2), 609-612 (2005)
The effects of surface treatments of the substrates on high-quality GaN crystal growth.
Fujimori T, et al.
Journal of Crystal Growth, 372, 73-77 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service