414069
Aluminum oxide
pellets, 3 mm
Synonym(s):
Alumina
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
form
pellets
particle size
3 mm
mp
2040 °C (lit.)
SMILES string
O=[Al]O[Al]=O
InChI
1S/2Al.3O
InChI key
TWNQGVIAIRXVLR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Aluminum oxide (alumina) is a common raw material for ceramic materials. It is a good electrical insulator that shows stability at high temperature and resistance to chemicals. Alumina is an important source for aluminum and has been prepared from high-alumina ladle furnace refining slag. It is reported to enhance the reaction rate of aldose to ketose conversion by Lobry de Bruyn–Alberda van Ekenstein transformation. It is a key constituent of NOx storage catalysts (NSC) which is employed to get rid of nitrogen oxides from the exhaust gas of lean burn engines. A study of the surface passivation characteristics of aluminium oxide on crystalline silicon shows many advantages which can be applied in silicon solar cell production.
Application
Aluminum oxide may be used as a dopant to produce optical fiber amplifiers and high-power fiber lasers.
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Study on the mechanism of the reaction of NO2 with aluminium oxide.
Apostolescu N, et al.
Applied Catalysis. B, Environmental, 51(1), 43-50 (2004)
High contrast laser marking of alumina.
Penide J, et al.
Applied Surface Science, 336, 118-128 (2015)
Alumina extraction from high-alumina ladle furnace refining slag.
Zhou Y, et al.
Hydrometallurgy, 140, 14-19 (2013)
Aldose-ketose interconversion in pyridine in the presence of aluminium oxide.
Ekeberg D, et al.
Carbohydrate Research, 342(14), 1992-1997 (2007)
Alumina as a dopant in optical fiber by OVD.
Wang J.
Applied Physics. A, Materials Science & Processing, 116(2), 505-518 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service