Skip to Content
Merck
All Photos(3)

Key Documents

34871

Sigma-Aldrich

1-Propanol

suitable for HPLC, ≥99.9%

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.05
Assay:
≥99.9%
technique(s):
HPLC: suitable
bp:
97 °C (lit.)
vapor pressure:
10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.1 (vs air)

Quality Level

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as C2H5COOH)
≤0.05% water (Karl Fischer)

evapn. residue

≤0.005%

transmittance

220 nm, ≥40%
240 nm, ≥85%
275 nm, ≥99%

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

λ

neat

UV absorption

λ: 220 nm Amax: ≤0.40
λ: 240 nm Amax: ≤0.071
λ: 275 nm Amax: ≤0.0044

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Propanol is a linear alcohol. It exhibits Debye-type relaxation process which ressembles the α-relaxation and Johari-Goldstein type β-relaxation features of the supercooled liquids (nonhydrogen-bonding type).[1] It has been tested as a substitute of the fuel for various fuel cells.[2] Its biosynthesis from glucose using Escherichia coli strain has been proposed.[3]

Application

1-Propanol may be employed as a solvent for the synthesis of mesoporous alumina.[4]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Juan Antonio Cecilia et al.
Materials (Basel, Switzerland), 11(9) (2018-09-12)
Vanadium oxide (V₂O₅) species has been supported on different porous clay heterostructures (with silica pillars, silica-zirconia with a molar ratio Si/Zr = 5 and silica-titania with a molar ratio Si/Ti = 5) by wetness incipient method. All catalysts were characterized
Mesoporous alumina (I): comparison of synthesis schemes using anionic, cationic, and non-ionic surfactants.
Ray JC, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 100(1), 183-190 (2007)
Dynamics of glass-forming liquids. III. Comparing the dielectric α-and β-relaxation of 1-propanol and o-terphenyl.
Hansen C, et al.
J. Chem. Phys. , 107, 1086-1093 (1997)
C R Shen et al.
Metabolic engineering, 10(6), 312-320 (2008-09-09)
Production of higher alcohols via the keto-acid intermediates found in microorganism's native amino-acid pathways has recently shown promising results. In this work, an Escherichia coli strain that produces 1-butanol and 1-propanol from glucose was constructed. The strain first converts glucose
Dieuwke Sevenster et al.
Science (New York, N.Y.), 339(6121), 830-833 (2013-02-16)
Although reconsolidation opens up new avenues to erase excessive fear memory, subtle boundary conditions put constraints on retrieval-induced plasticity. Reconsolidation may only take place when memory reactivation involves an experience that engages new learning (prediction error). Thus far, it has

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service