Skip to Content
Merck
All Photos(3)

Key Documents

319929

Sigma-Aldrich

1,2-Dichloroethane

ACS reagent, ≥99.0%

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
EC Number:
MDL number:
UNSPSC Code:
12352101
eCl@ss:
39050106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

impurities

≤0.0003 meq/g Titr. acid
≤0.03% water

evapn. residue

≤0.002%

color

APHA: ≤10
clear

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloroethane (ethylene dichloride, DCE) is a solvent. It is one of the degradation by-product released from vinyl chloride manufacturing industries (EDC-tar). Its mutagenic activity has been tested on Salmonella typhimurium TA 1535. Its catalytic degradation over titania/silica (acid catalysts) has been investigated.

Application

1,2-Dichloroethane (ethylene dichloride, DCE) may be used in the following studies:
  • As electron acceptor in the culture medium of Dehalococcoides ethenogenes 195.
  • As model compound for chlorinated organics in various studies.
  • Chemical doping during the fabrication of Cl-doped few-layer WS2 or MoS2 back gate field-effect transistors (FETs).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingming Yang et al.
Nano letters, 14(11), 6275-6280 (2014-10-14)
Low-resistivity metal-semiconductor (M-S) contact is one of the urgent challenges in the research of 2D transition metal dichalcogenides (TMDs). Here, we report a chloride molecular doping technique which greatly reduces the contact resistance (Rc) in the few-layer WS2 and MoS2.
X Maymó-Gatell et al.
Applied and environmental microbiology, 65(7), 3108-3113 (1999-07-02)
"Dehalococcoides ethenogenes" 195 can reductively dechlorinate tetrachloroethene (PCE) completely to ethene (ETH). When PCE-grown strain 195 was transferred (2% [vol/vol] inoculum) into growth medium amended with trichloroethene (TCE), cis-dichloroethene (DCE), 1,1-DCE, or 1,2-dichloroethane (DCA) as an electron acceptor, these chlorinated
Decomposition of 1, 2-dichloroethane on titanium dioxide/silica.
Imamura S, et al.
Industrial & Engineering Chemistry Research, 28(10), 1449-1452 (1989)
U Rannug et al.
Chemico-biological interactions, 20(1), 1-16 (1978-01-01)
One of the main components in the waste products from vinyl chloride industries (EDC-tar), is ethylene dichloride (1,2-dichloroethane). This compound has been tested for mutagenicity on Salmonella typhimurium TA 1535. It is concluded that 1,2-dichloroethane gives a weak direct mutagenic
Tatiana Wolfe et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(5), 1277-1283 (2015-02-06)
We report potent radiosensitization of prostate cancers in vitro and in vivo using goserelin-conjugated gold nanorods. Progressive receptor-mediated internalization of conjugated nanorods over time increases the radiation interaction cross-section of cells and contributes to the effects observed in vitro. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service