Skip to Content
Merck
All Photos(2)

Key Documents

311383

Sigma-Aldrich

Arsenic(III) oxide

ACS reagent (primary standard)

Synonym(s):

Arsenic trioxide, Arsenous acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As2O3
CAS Number:
Molecular Weight:
197.84
EC Number:
MDL number:
UNSPSC Code:
12171602
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent (primary standard)
form:
powder
solubility:
dilute HCl: insoluble ≤0.01%

grade

ACS reagent (primary standard)

Quality Level

vapor pressure

0 hPa ( 66 °C)

Assay

99.95-100.05%

form

powder

reaction suitability

reagent type: catalyst
core: arsenic

ign. residue

≤0.02%

solubility

dilute HCl: insoluble ≤0.01%

anion traces

S2-: passes test (lim. ~0.001%)
chloride (Cl-): ≤0.005%

cation traces

Fe: ≤5 ppm
Pb: ≤0.002%
Sb: ≤0.05%

SMILES string

O=[As]O[As]=O

InChI

1S/As2O3/c3-1-5-2-4

InChI key

IKWTVSLWAPBBKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arsenic (III) oxide, also referred to as arsenic trioxide, is an amphoteric oxide that is used as a raw material to produce several arsenic-based compounds.

Application

Arsenic (III) oxide is used
  • In the synthesis of arsenic pentafluoride (AsF5) by static fluorination method in a closed system.
  • In the preparation of arsenic-doped ZnO films by adding As2O3 to zinc acetate, 2-methoxyethanol, and monoethanolamine by sol-gel spin coating method.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Respiratory system,Cardio-vascular system,Gastrointestinal tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Liu et al.
Science China. Life sciences, 63(11), 1744-1754 (2020-05-10)
This study aimed to explore macrophage polarization in acute graft-versus-host disease after hematopoietic stem cell transplantation, and investigated if arsenic trioxide (ATO) could correct this imbalance. In the colon of GVHD mice, we found that the number of F4/80+iNOS+ cells
Yanfei Jia et al.
PloS one, 8(1), e54774-e54774 (2013-02-06)
Alpha-fetoprotein (AFP)-producing gastric cancer (AFPGC), represented by the production of AFP, has a more aggressive behavior than common gastric cancer. The underlying mechanisms are not well understood. Arsenic trioxide (As(2)O(3)) is used clinically to treat acute promyelocytic leukemia(APL) and has
Yuhua Fan et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 31(1), 80-91 (2013-02-01)
Arsenic trioxide (As2O3) is a highly effective agent for treatment of acute promyelocytic leukemia (APL). However, consecutively administered As2O3 induces serious adverse cardiac effects, including long QT syndrome (LQTs) and even sudden cardiac death. Previous studies have shown that genistein
Jianing Wu et al.
Toxicology letters, 220(1), 61-69 (2013-04-02)
Notch signaling has been demonstrated to have a central role in cancer stem-like cells (CSLCs) in glioblastoma multiforme (GBM). We have recently demonstrated the inhibitory effect of arsenic trioxide (ATO) on CSLCs in glioblastoma cell lines. In this study we
Michael T Brady et al.
Leukemia research, 37(7), 822-828 (2013-05-01)
Signal transducer and activator of transcription (STAT) 3 inhibits dendritic cell (DC) differentiation and is constitutively activated in blasts of approximately half of AML patients. We investigated the correlation between STAT3 activity, DC maturation and the ability to stimulate T-cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service