Skip to Content
Merck
All Photos(1)

Key Documents

31107

Sigma-Aldrich

Ammonium chloride

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050207
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
form:
crystalline powder

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

Assay

≥99.5%

form

crystalline powder

pH

4.5-5.5 (20 °C, 5%)

mp

340 °C (subl.) (lit.)

anion traces

nitrate (NO3-): ≤5 mg/kg
phosphate (PO43-): ≤2 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤5 mg/kg
Cu: ≤2 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤2 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ammonium chloride can be used as a catalyst:
  • To synthesize 3,4-dihydropyrimidin-2-(1H)-ones via solvent-free Biginelli-condensation reaction.
  • For the aldol condensation of carbonyl compounds.
  • In the thia-Michael addition reaction of thiols to different α-enones.
It can also be used as:
  • A promoter in the enantioselective hydrogenation of an unprotected β-enamine amides.
  • A pH-adjuster in the preparation of waterborne glucose silicone resin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of ammonium chloride as an effective promoter of the asymmetric hydrogenation of a beta-enamine amide
Clausen A, et al.
Organic Process Research & Development, 10, 723-726 (2006)
Ammonium chloride-catalyzed one-pot synthesis of 3, 4-dihydropyrimidin-2-(1H)-ones under solvent-free conditions
Shaabani A, et al.
Tetrahedron Letters, 44, 857-859 (2003)
Physical and mechanical properties of poplar wood modified by glucose-urea-melamine resin/sodium silicate compound
Liu Q, et al.
Forests, 12, 127-127 (2021)
Ammonium chloride-catalyzed carbon-sulfur bond formation in water
Chen W, et al.
Catalysis Communications, 9, 1079-1081 (2008)
The Vibrational Spectra of Molecules and Complex Ions in Crystals III. Ammonium Chloride and Deutero-Ammonium Chloride.
Wagner EL and Hornig DF.
J. Chem. Phys. , 18(3), 296-304 (1950)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service