Skip to Content
Merck
All Photos(2)

Key Documents

310433

Sigma-Aldrich

Copper(II) oxide

needles, mixture of CuO and Cu2O, ACS reagent

Synonym(s):

Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
needles
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

form

needles

reaction suitability

reagent type: catalyst
core: copper

impurities

≤0.002% C compounds
≤0.002% N compounds

anion traces

sulfate (SO42-): ≤0.012%

cation traces

C: ≤0.002%
N: ≤0.002%

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effects of Copper(II) Oxide on the Co-Pyrolysis of Waste Polyester Enameled Wires and Poly(vinyl chloride).: This research demonstrates the role of Copper(II) oxide in enhancing the co-pyrolysis process of waste materials, contributing to environmental sustainability and waste management solutions in analytical chemistry (Zhu G et al., 2023).

  • Adsorption isotherms and kinetics for Pb(ii) ion removal from aqueous solutions with biogenic metal oxide nanoparticles.: The study explores the efficiency of Copper(II) oxide nanoparticles in removing lead ions from water, highlighting its potential for pollution control and water treatment technologies (Sadyrbekov DT et al., 2023).

  • Preparation of surface-modified electrode of copper(ii) oxide mixed with the molecularly imprinted polymer for enhancement of melamine detection with photoelectrochemical technique.: This article describes the use of Copper(II) oxide in developing advanced sensors for detecting contaminants like melamine, demonstrating its application in food safety and public health (Thiwawong T et al., 2023).

  • Synthesis and characterization of copper oxide nanoparticles: its influence on corn (Z. mays) and wheat (Triticum aestivum) plants by inoculation of Bacillus subtilis.: This research investigates the impact of Copper(II) oxide nanoparticles on plant growth, providing insights into agricultural applications and plant biology (Sharma R et al., 2023).


Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A refinement of the crystal structure of copper (II) oxide with a discussion of some exceptional esd's.
?sbrink S and Norrby LJ.
Acta Crystallographica Section B, Structural Science, 26(1), 8-15 (1970)
DSC kinetics of the thermal decomposition of copper (II) oxalate by isoconversional and maximum rate (peak) methods.
Muraleedharan K and Kripa S.
Journal of Thermal Analysis and Calorimetry, 115(2), 1969-1978 (2014)
Copper (II) oxide nanowires for p-type conductometric NH3 sensing.
Shao F, et al.
Applied Surface Science, 311, 177-181 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 523-523 (1994)
Wei Jian Foo et al.
Nanoscale, 5(2), 759-764 (2012-12-12)
Here we have demonstrated the preparation of high-quality, monodispersed and tunable phases of Cu nanoparticles. Structural and chemical composition studies depict the evolution of Cu-Cu(2)O-CuO nanoparticles at various process stages. The loading of Cu and Cu oxide nanoparticles on TiO(2)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service