238988
Drierite™
with indicator, 8 mesh
Synonym(s):
Calcium sulfate, Gypsum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Recommended Products
Quality Level
form
chunks
particle size
8 mesh
SMILES string
[Ca++].[O-]S([O-])(=O)=O
InChI
1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Drierite™ can be used as a desiccant in the:
- Dinuclear nickel-catalyzed asymmetric vinylogous Mannich-type reaction of α, β-unsaturated γ-butyrolactam and N-boc imines to synthesize vinylogous Mannich adducts.
- Catalytic epoxidation of propylene glycol acetates to propylene oxide.
- Rare earth-alkali metal heterobimetallic complex-catalyzed enantioselective Aza-Michael addition of alkoxylamines to α, β-unsaturated carbonyl compounds to synthesize β-amino carbonyl compounds.
Legal Information
Drierite is a trademark of W.A. Hammond Drierite Co., LTD
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Repr. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yan Bao et al.
Plant methods, 16, 126-126 (2020-09-25)
Tocochromanols are a group of lipid-soluble antioxidants produced by all plants and include the tocopherols, which are ubiquitous in the plant kingdom, and the biosynthetically-related compounds tocotrienols, which along with tocopherols commonly occur in seed of monocots. Most tocochromanols have
The ontogeny of personality in the wild guinea pig.
Guenther A, et al.
Animal behaviour, 90, 131-139 (2014)
Enthalpies of vaporization of organic compounds.
Howard PB and Wadso I.
Acta Chemica Scandinavica, 24, 145-149 (1970)
The analysis of carbon monoxide in urban air at the ppm level, and the normal carbon monoxide value.
L Dubois et al.
Journal of the Air Pollution Control Association, 16(3), 135-139 (1966-03-01)
Respirometry: anhydrous drierite equilibrates with carbon dioxide and increases washout times.
White CR, et al.
Physiological and Biochemical Zoology : Pbz, 79(5), 977-988 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service