Skip to Content
Merck
All Photos(3)

Key Documents

237108

Sigma-Aldrich

Barium carbonate

ACS reagent, ≥99%

Synonym(s):

Witherite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCO3
CAS Number:
Molecular Weight:
197.34
Beilstein:
7045119
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
99.0-101.0% (ACS specification)
grade:
ACS reagent
form:
powder or chunks

grade

ACS reagent

Quality Level

Assay

≥99%
99.0-101.0% (ACS specification)

form

powder or chunks

impurities

≤0.002 meq/g water-soluble titr. base
≤0.005% oxidizers
≤0.015% insol. dil. HCl

pH

6.8 (37 °C, 3.67 g/L)

anion traces

S2-: ≤0.001%
chloride (Cl-): ≤0.002%

cation traces

Ca: ≤0.05%
Fe: ≤0.002%
K: ≤0.005%
Na: ≤0.02%
Sr: ≤0.7%
heavy metals (as Pb): ≤0.001%

SMILES string

[Ba++].[O-]C([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium carbonate (BaCO3) is an important alkaline-earth metal material in the production of barium salts, barium ferrite, pigments, ceramics, optical glass, and electric condensers. Additionally, it serves as a precursor to produce superconductors and ceramic materials.

Application

Barium carbonate is used:
  • In the preparation of barium titanate using a mixture of BaCO3 and titania (TiO2) by solid-state reaction.
  • As an additive in the preparation of NBC (neodymium – barium –copper) ceramics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of anatase and rutile with barium carbonate.
AMIN A, et al.
Journal of the American Ceramic Society. American Ceramic Society, 66(10), 733-738 (1983)
Ali Khorsand Zak et al.
Nanoscale research letters, 9(1), 399-399 (2014-09-02)
Pure zinc oxide and zinc oxide/barium carbonate nanoparticles (ZnO-NPs and ZB-NPs) were synthesized by the sol-gel method. The prepared powders were characterized by X-ray diffraction (XRD), ultraviolet-visible (UV-Vis), Auger spectroscopy, and transmission electron microscopy (TEM). The XRD result showed that
Characterization of barium titanate powders: barium carbonate identification.
Lopez MCB, et al.
Journal of the American Chemical Society, 82(7), 1777-1786 (1999)
D Ekeberg et al.
Carbohydrate research, 335(2), 141-146 (2001-09-25)
A simple, but low-yielding method for the synthesis of 3-hexuloses has been elaborated. Oxidation of 1,2:5,6-di-O-isopropylidenehexitols with bromine in the presence of barium carbonate, followed by mild-acid hydrolysis of the oxidation products gave the free hexuloses. Oxidation occurred at only
Xiaorang Tian et al.
Journal of hazardous materials, 160(2-3), 589-593 (2008-04-25)
In this study, the exploration of making acidproof fracturing proppants using red mud was carried out. The main raw materials are red mud and the refractory waste. During the exploration, three methods were explored to enhance the acid resistance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service