All Photos(3)
About This Item
Linear Formula:
LiNO3
CAS Number:
Molecular Weight:
68.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
form:
powder or crystals
solubility:
water: soluble(lit.)
Recommended Products
Quality Level
product line
ReagentPlus®
form
powder or crystals
concentration
9-11% Li (ICP)
pH
7-9 (20 °C, 50 g/L)
mp
264 °C (lit.)
solubility
water: soluble(lit.)
SMILES string
[Li+].[O-][N+]([O-])=O
InChI
1S/Li.NO3/c;2-1(3)4/q+1;-1
InChI key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Lithium nitrate (LiNO3) is an inorganic compound used as an electrolyte additive in rechargeable Li-S (lithium-sulfur) batteries. LiNO3 acts as an oxidizing agent.
Application
Lithium nitrate is used:
- As an electrolyte and redox mediator in the Li-O2 battery.
- In the preparation of LiNO3/natural phosphate (NP) catalyst. This LiNO3/NP catalyst can be employed in the synthesis of chalcones.
- As a regenerable SEI (solid–electrolyte interphase) stabilizing agent in the rechargeable Li/O2 batteries.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Vapor-liquid equilibrium of ammonia+ lithium nitrate+ water and ammonia+ lithium nitrate solutions from (293.15 to 353.15) K.
Libotean S, et al.
Journal of Chemical and Engineering Data, 52(3), 1050-1055 (2007)
Dramatic activity enhancement of natural phosphate catalyst by lithium nitrate. An efficient synthesis of chalcones.
Sebti S, et al.
Catalysis Communications, 3(8), 335-339 (2002)
Wesley Walker et al.
Journal of the American Chemical Society, 135(6), 2076-2079 (2013-01-31)
A major challenge in the development of rechargeable Li-O(2) batteries is the identification of electrolyte materials that are stable in the operating environment of the O(2) electrode. Straight-chain alkyl amides are one of the few classes of polar, aprotic solvents
Eagleson M.
Concise Encyclopedia Chemistry, 605-605 (1994)
Sung-Ho Shin et al.
ACS nano, 8(10), 10844-10850 (2014-09-30)
We present a method to develop high performance flexible piezoelectric nanogenerators (NGs) by employing Li-doped ZnO nanowires (NWs). We synthesized Li-doped ZnO NWs and adopted them to replace intrinsic ZnO NWs with a relatively low piezoelectric coefficient. When we exploited
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service