Skip to Content
Merck
All Photos(2)

Key Documents

222283

Sigma-Aldrich

Magnesium perchlorate

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2
CAS Number:
Molecular Weight:
223.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
flakes
powder, chunks or granules

grade

ACS reagent

Quality Level

form

flakes
powder, chunks or granules

reaction suitability

reagent type: oxidant

concentration

>10% Mg (EDTA titration)

impurities

≤0.005 meq/g Titr. free acid
≤0.025 meq/g Titr. base

loss

≤8% loss on drying

suitability

passes test for moisture absorption

SMILES string

[Mg++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Mg/c2*2-1(3,4)5;/h2*(H,2,3,4,5);/q;;+2/p-2

InChI key

MPCRDALPQLDDFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium perchlorate (Mg(ClO4)2) is widely used as drying agent for gases. It can remove water from gases (with no organic contaminants) at the rate of 0.001mgwater/l.

Application

Magnesium perchlorate (Mg(ClO4)2) may be employed as a catalyst in the following studies:
  • Preparation of α-aminophosphonates.
  • Enantioselective Diels-Alder reaction between cyclopentadiene and 3-acryloyl-1,3-oxazolin-2-one.
  • Preparation of imines and phenylhydrazones.
  • Protection of alcohols in the form of t-butyl ethers.
Magnesium perchlorate may be used as a catalyst in the synthesis of the following compounds:
  • α-Aminophosphonates via three-component reaction between an amine, an aldehyde or a ketone and a di-/trialkyl phosphite.
  • Imines and phenylhydrazones by the condensation of carbonyl compounds with amines and phenylhydrazine.
  • Knoevenagel adducts via Knoevenagel condensation between β-diketones and aliphatic or aromatic aldehydes.
It may also be used to catalyze the protection of alcohols as t-butyl ethers.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium perchlorate as efficient Lewis acid for the Knoevenagel condensation between ?-diketones and aldehydes.
Bartoli G, et al.
Tetrahedron Letters, 49(16), 2555-2557 (2008)
Giuseppe Bartoli et al.
Organic letters, 7(3), 427-430 (2005-01-28)
[reaction: see text] A new mild method for protecting alcohols as t-butyl ethers is reported. The reaction proceeds with Mg(ClO4)2 and Boc2O and shows general applicability. The deprotection of t-butyl ethers has also been revisited. Preliminary results indicate the CeCl3
Magnesium perchlorate as an efficient catalyst for the synthesis of imines and phenylhydrazones.
Chakraborti AK, et al.
Tetrahedron Letters, 45(41), 7641-7644 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 343-343 (1994)
The first enantioselective synthesis of both Diels-Alder enantiomers with the same bis (oxazoline)-magnesium perchlorate chiral catalyst.
Desimoni G, et al.
Tetrahedron Letters, 37(17), 3027-3030 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service