Skip to Content
Merck
All Photos(1)

Key Documents

22152

Sigma-Aldrich

Celite® 512 medium

filter aid, calcined

Synonym(s):

Diatomaceous earth, calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.54

Quality Level

form

powder

quality

calcined

composition

Al2O3, 4.0%
CaO, 0.6%
Fe2O3, 1.1%
Na2O + K2O, 1.1%
SiO2, 91.5%

median particle size

16.4 μm (acc. to granulometric method (laser sizer))

pH

<7 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Celite® 512 medium is a filter aid derived from diatomaceous earth. Diatomaceous earth is made up of natural amorphous silica formed from the fossilized skeletons of diatoms. It is extracted, crushed, dried and calcined with flux material. DE is widely used as filler in construction materials, paints and as an anti-caking agent for agricultural chemicals. It has high water and oil absorption ability.

Caution

Not Intended for cGMP Parenteral Pharmaceutical Manufacturing

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian R McMinn et al.
Journal of virological methods, 179(2), 423-428 (2011-12-06)
The effective recovery of adenovirus from water is a critical first step in developing a virus occurrence method able to provide accurate data for risk assessments and other applications. During virus concentration, electropositive filters are typically eluted with beef extract
Manipulating surface wettability and oil absorbency of diatomite depending on processing and ambient conditions.
Ozen I, et al.
Applied Surface Science, 332, 22-31 (2015)
H Checkoway et al.
British journal of industrial medicine, 50(7), 586-597 (1993-07-01)
A cohort mortality study was conducted among workers from two plants in the diatomaceous earth mining and processing industry in California. Diatomaceous earth consists of the skeletal remains of diatoms. Exposure to amorphous (non-crystalline) and crystalline silica in the form
Federica Maraschi et al.
Analytical biochemistry, 426(2), 142-146 (2012-05-01)
This article presents the first results demonstrating that total silicon trace concentration in human ventricular whole blood may be used as a further marker in the diagnosis of drowning. The difference in silicon content between the left and right ventricles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service