Skip to Content
Merck
All Photos(8)

Key Documents

221473

Sigma-Aldrich

Potassium hydroxide

Potassium hydroxide

ACS reagent, ≥85%, pellets

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
pellets
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

suitable for EPA 1621
suitable for EPA 1633

vapor pressure

1 mmHg ( 719 °C)

Assay

≥85%

form

pellets

impurities

≤0.001% N compounds
≤2.0% K2CO3
10-15% water

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
Mg: ≤0.002%
Na: ≤0.05%
Ni: ≤0.001%
heavy metals: ≤0.001% (by ICP)

application(s)

PFAS testing

storage temp.

room temp

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide is an inorganic salt mainly used as a precursor to synthesize other potassium compounds such as potassium acetate,[1] potassium bicarbonate and potassium carbonate.[2] On a commercial scale, it is synthesized by the electrolysis of potassium chloride.[3]

Application

Potassium hydroxide may be used in the following processes:
  • Conversion of nitriles to amide in tert-butanol.[4]
  • Cross-coupling of aryl halides with different sulfur-, oxygen- and nitrogen-based nucleophiles in dimethyl sulfoxide.[5]
  • As an activator for the preparation of activated carbons from corn cobs.[6]
  • As a base for the N-alkylation of nitrogen heterocycles in polyethylene glycol-ether solvents.[7]
  • Synthesis of (S)-1-(4-benzyl-2-thioxothiazolidin-3-yl)propan-1-one from (S)-phenylalaninol.[8]
  • To catalyze the polymerization of propylene oxide.[9]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Alkylation of pyrrole, indole, and several other nitrogen heterocycles using potassium hydroxide as a base in the presence of polyethylene glycols or their dialkyl ethers
Sukata K
Bulletin of the Chemical Society of Japan, 56(1), 280-284 (1983)
Preparation of high surface area activated carbon from corn by chemical activation using potassium hydroxide
Bagheri N & Abedi J
Chemical Engineering Research and Design, 87(8), 1059-1064 (2009)
A study of the polymerization of propylene oxide catalyzed by anhydrous potassium hydroxide
Steiner EC, et al
Journal of the American Chemical Society, 86(21), 4678-4686 (1964)
Dimethyl Sulfoxide/Potassium Hydroxide: A Superbase for the Transition Metal-Free Preparation of Cross-Coupling Products
Yuan Y, et al
Advanced Synthesis & Catalysis, 352(17), 2892-2898 (2010)
Potassium acetate by fermentation with Clostridium thermoaceticum
Shah MM, et al.
Applied Biochemistry and Biotechnology (1997)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service