Skip to Content
Merck
All Photos(1)

Key Documents

Z3000000

Zopiclone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Methyl-1-piperazinecarboxylic acid 6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, Imovane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClN6O3
CAS Number:
Molecular Weight:
388.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

zopiclone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4

InChI

1S/C17H17ClN6O3/c1-22-6-8-23(9-7-22)17(26)27-16-14-13(19-4-5-20-14)15(25)24(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3

InChI key

GBBSUAFBMRNDJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Zopiclone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Hajak et al.
PharmacoEconomics, 10 Suppl 1, 29-38 (1995-12-09)
Insomnia, especially when chronic, is associated with disturbances in daytime well-being and performance, resulting in a poor quality of life for those affected. Zopiclone has been proven as a drug that favourably balances sleep induction and maintenance as well as
A N Wadworth et al.
Drugs & aging, 3(5), 441-459 (1993-09-01)
Zopiclone is a cyclopyrrolone which is chemically unrelated to the benzodiazepines and is thought to act on the GABAA receptor complex at a site distinct from, but closely related to, the benzodiazepine binding site. The hypnotic efficacy of zopiclone administered
J G Bramness et al.
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 118(13), 2029-2032 (1998-07-10)
In 1994 zopiclone (Imovane), a cyclopyrrolon, was introduced in Norway as a new kind of hypnotic. In 1996 zopiclone had a 26% share of the hypnotic market. This review of relevant literature has revealed a lack of documentation on the
B Musch et al.
International clinical psychopharmacology, 5 Suppl 2, 147-158 (1990-04-01)
Effective and safe hypnotics exist especially since the introduction of benzodiazepines (BZD) which appeared to bring major advantages over barbiturates. Ideally a new hypnotic should induce and maintain sleep without producing residual effect during the day and should be devoid
Joris C Verster et al.
Current drug safety, 6(4), 209-218 (2011-12-02)
Zopiclone (7.5 mg) is frequently used as a positive control in studies that examine the residual effects of hypnotic drugs on driving ability and related skills. This review summarizes studies examining the effects of zopiclone, and discusses its usefulness as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service