Skip to Content
Merck
All Photos(1)

Key Documents

Y0001907

Isopropyl isostearate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Isopropyl 16-methylheptadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H42O2
CAS Number:
Molecular Weight:
326.56
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isopropyl isostearate

manufacturer/tradename

EDQM

format

neat

storage temp.

2-8°C

InChI

1S/C21H42O2/c1-19(2)17-15-13-11-9-7-5-6-8-10-12-14-16-18-21(22)23-20(3)4/h19-20H,5-18H2,1-4H3

InChI key

NEOZOXKVMDBOSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isopropyl isostearate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

344.3 °F

Flash Point(C)

173.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moisturizers: what they are and a practical approach to product selection.
Kraft J N, et al.
Skin Therapy Letter, 10(5), 1-8 (2005)
Final report on the safety assessment of isopropyl isostearate.
Andersen FA.
Journal of the American College of Toxicology, 11(1), 43-49 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service