Skip to Content
Merck
All Photos(1)

Key Documents

Y0001615

Beta-sitosterol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

β-Sitosterol, α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Beilstein:
1916165
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

beta-sitosterol

manufacturer/tradename

EDQM

mp

136-140 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@H](C(C)C)CC

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Beta-sitosterol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T J Wilt et al.
BJU international, 83(9), 976-983 (1999-06-15)
To conduct a systematic review of the evidence for the efficacy of beta-sitosterol in men with symptomatic benign prostatic hyperplasia (BPH). Studies were identified through Medlinetrade mark (1966-98), EMBASEtrade mark, Phytodok, the Cochrane Library, bibliographies of identified trials and review
Natalia Toro-Funes et al.
Journal of agricultural and food chemistry, 62(39), 9539-9543 (2014-09-05)
Ultra-high-pressure homogenization (UHPH) is an emerging technology based on the dynamic application of high pressure to obtain safe and high-quality liquid foods. The effect of six UHPH treatments at 200 and 300 MPa with different inlet temperatures (T(in)) (55, 65
Z Ovesná et al.
Neoplasma, 51(6), 407-414 (2005-01-11)
Substantial attention has been given to primary cancer prevention in daily life. Dietary factors are through to contribute to as much as one-third of the factors influencing the development of cancer. Ones of the components of a plant-based diet are
Na-Ra Han et al.
Experimental biology and medicine (Maywood, N.J.), 239(4), 454-464 (2014-02-11)
The compound β-sitosterol (BS) is one of the most common forms of phytosterols and has anti-cancer, anti-oxidant, anti-bacterial, and anti-inflammatory effects. However, the effect of BS on atopic dermatitis (AD) has not been elucidated. Therefore, we investigated whether BS would
Helena E Miettinen et al.
Journal of lipid research, 55(12), 2644-2654 (2014-10-11)
We examined serum cholesterol synthesis and absorption markers and their association with neonatal birth weight in obese pregnancies affected by gestational diabetes mellitus (GDM). Pregnant women at risk for GDM (BMI >30 kg/m²) were enrolled from maternity clinics in Finland.

Protocols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service