Skip to Content
Merck
All Photos(1)

Key Documents

Y0001442

N-[(cis-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-[[cis-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nateglinide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

N-[(cis-4-Isopropylcyclohexyl)carbonyl]-ᴅ-phenylalanine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Phielix et al.
Diabetes, obesity & metabolism, 15(10), 915-922 (2013-04-12)
Thiazoledinediones decrease blood glucose by their insulin-sensitizing properties. Here, we examined whether pioglitazone plus nateglinide (PIO) interferes with hepatocellular lipid (HCL) content and/or improves insulin sensitivity in well-controlled non-obese patients with type 2 diabetes mellitus (T2DM). Sixteen patients [body mass
F Zheng et al.
Journal of endocrinological investigation, 36(7), 489-496 (2013-01-18)
Recent studies highlight an important role of ghrelin in glucose homeostasis, while the association between ghrelin regulation and glucose fluctuation is unclear. We compared the effects of two postprandial hypoglycemic agents on ghrelin response and determined the contribution of ghrelin
Ranee Chatterjee et al.
American journal of hypertension, 26(6), 723-726 (2013-02-19)
Low and low-normal serum potassium is associated with an increased risk of diabetes. We hypothesized that the protective effect of valsartan on diabetes risk could be mediated by its effect of raising serum potassium. We analyzed data from the Nateglinide
Subhash S Vaghani et al.
Current drug delivery, 9(6), 628-636 (2012-03-29)
In current research, chitosan was reacted with mono-chloroacetic acid under alkaline condition to prepare carboxymethyl chitosan (CMCTs). The degree of substitution (Ds) on prepared CMCTs was found to be 0.68. CMCTs was used as a potential carrier for pH specific
Jin-Kui Yang et al.
Clinical drug investigation, 33(3), 185-191 (2013-01-23)
Diabetes mellitus has become a major public health problem in China. This open-label, prospective, multicentre, post-marketing surveillance study was conducted to investigate the efficacy and safety of nateglinide in combination with metformin in Chinese patients with type 2 diabetes (T2DM).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service