Skip to Content
Merck
All Photos(1)

Key Documents

Y0001413

Fulvestrant for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fulvestrant, (7α,17β)-7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol, Faslodex, ICI 182,780, ZD 182780, ZD 9238, ZM 182780

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H47F5O3S
CAS Number:
Molecular Weight:
606.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fulvestrant

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)Cc4cc(O)ccc24

InChI

1S/C32H47F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,21-22,26-29,38-39H,2-10,13-20H2,1H3/t22-,26-,27+,28+,29-,30+,41?/m1/s1

InChI key

VWUXBMIQPBEWFH-WCCTWKNTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fulvestrant for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Fulvestrant (ICI 182,780) is a selective estrogen receptor down-regulator (SERD). Fulvestrant is a high affinity estrogen receptor antagonist. IC50 = 0.29 nM. Fulvestrant is the first "pure" antiestrogen with no agonistic activity both in vitro and in vivo.
Fulvestrant is a selective estrogen receptor down-regulator (SERD); high affinity estrogen receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Xin et al.
Environmental science & technology, 53(14), 8371-8380 (2019-06-30)
As alternatives to perfluorooctanoic acid (PFOA), hexafluoropropylene oxide (HFPO) homologues, including hexafluoropropylene oxide dimer acid (HFPO-DA), hexafluoropropylene oxide trimer acid (HFPO-TA), and hexafluoropropylene oxide tetramer acid (HFPO-TeA), have been used in the fluoropolymer industry for a long period of time.
L Estévez et al.
Cancer treatment reviews, 39(2), 136-141 (2012-07-17)
Fulvestrant is a selective estrogen receptor downregulator, behaving as a complete antagonist. It was initially approved, at a dose of 250 mg, to treat hormone dependant breast cancer in second line setting. However, a series of pharmacological and pre-clinical studies
Is fulvestrant ("Faslodex") just another selective estrogen receptor modulator?
A Howell
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society, 16 Suppl 2, 521-523 (2006-10-03)
Peter Kabos et al.
Expert opinion on pharmacotherapy, 11(5), 807-816 (2010-02-16)
The role of estrogen deprivation for the treatment of breast cancer has been understood since the 1800s. Pharmacologic advances in the field in the past decades, including tamoxifen and the aromatase inhibitors, have contributed significantly to the reduced mortality of
Stephen Chia et al.
Breast (Edinburgh, Scotland), 17 Suppl 3, S16-S21 (2008-03-21)
With the aromatase inhibitors (AIs) replacing tamoxifen as the first-line treatment for postmenopausal women with hormone receptor-positive early and advanced breast cancer, there is a need to evaluate appropriate endocrine treatment options following AI failure. However, until recently, there were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service