Skip to Content
Merck
All Photos(1)

Key Documents

Y0001390

Cyproterone acetate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cyproterone acetate, 6-Chloro-1β,2β-dihydro-17-hydroxy-3′H-cyclopropa(1,2)-pregna-1,4,6-triene-3,20-dione acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29ClO4
CAS Number:
Molecular Weight:
416.94
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cyproterone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@](OC(C)=O)(C(C)=O)[C@@]1(C)CC[C@@]3([H])[C@@]2([H])C=C(Cl)C4=CC(=O)[C@@H]5C[C@@H]5[C@]34C

InChI

1S/C24H29ClO4/c1-12(26)24(29-13(2)27)8-6-16-14-10-20(25)19-11-21(28)15-9-18(15)23(19,4)17(14)5-7-22(16,24)3/h10-11,14-18H,5-9H2,1-4H3/t14-,15+,16-,17-,18-,22-,23-,24-/m0/s1

InChI key

UWFYSQMTEOIJJG-FDTZYFLXSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyproterone acetate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyproterone acetate--mechanism of action and clinical effectiveness in prostate cancer treatment.
F H Schröder
Cancer, 72(12 Suppl), 3810-3815 (1993-12-15)
Z Laron et al.
Journal of pediatric endocrinology & metabolism : JPEM, 13 Suppl 1, 805-810 (2000-09-02)
The authors review their experience (1967-present) in the use of cyproterone acetate (CPA) in precocious puberty. CPA was found effective in persistently suppressing pituitary gonadotropic secretion when administered orally at a dose of 50 mg b.i.d. (70-100 mg/d). After the
The antiandrogen cyproterone acetate: discovery, chemistry, basic pharmacology, clinical use and tool in basic research.
F Neumann
Experimental and clinical endocrinology, 102(1), 1-32 (1994-01-01)
A J Cooper et al.
Canadian journal of psychiatry. Revue canadienne de psychiatrie, 37(1), 33-39 (1992-02-01)
This study reports the short term effects in five pedophiles of the antiandrogenic drug cyproterone acetate (CPA) on nocturnal penile tumescence (NPT); penile responses to erotic stimuli in the laboratory; and sex hormones (testosterone, LH, FSH and prolactin). During the
T Rabe et al.
Drug safety, 14(1), 25-38 (1996-01-01)
The preclinical safety assessment of cyproterone acetate (CPA) with regard to liver tumorigenesis was based on tumorigenicity studies, which revealed no mutagenic potential. Recently, in vitro studies on the formation of adducts and the enhancement of DNA repair synthesis with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service