Skip to Content
Merck
All Photos(1)

Key Documents

Y0001139

Epinastine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9,13b-Dihydro-1H-dibenz[cf]imidazo[1,5-a]azepine hydrochloride, WAL-801Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3 · HCl
CAS Number:
Molecular Weight:
285.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

epinastine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].NC1=NCC2N1c3ccccc3Cc4ccccc24

InChI

1S/C16H15N3.ClH/c17-16-18-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)19(15)16;/h1-8,15H,9-10H2,(H2,17,18);1H

InChI key

VKXSGUIOOQPGAF-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Epinastine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Epinastine hydrochloride is a non-sedating H1 histamine receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shilpi Pradhan et al.
Expert opinion on drug metabolism & toxicology, 5(9), 1135-1140 (2009-07-28)
Epinastine is a topical ophthalmic antihistamine and a mast cell stabilizer indicated for treatment of symptoms associated with allergic conjunctivitis. The objective of this review is to provide a comprehensive overview of the pharmacology, clinical efficacy, tolerability and safety of
Masanori Ito et al.
International journal of pharmaceutics, 441(1-2), 121-127 (2012-12-19)
The study objective was to quantitatively predict a drug's bitterness and estimate bitterness masking efficiency using an electronic tongue (e-Tongue). To verify the predicted bitterness by e-Tongue, actual bitterness scores were determined by human sensory testing. In the first study
Takeo Tsujii et al.
Psychopharmacology, 207(1), 127-132 (2009-08-26)
Antagonists of histamine H(1) receptors (antihistamines) are widely used for the treatment of allergic disorders in children. These drugs' sedative effect on brain function, however, has been mostly examined in adults. The objective of this study was to examine the
Takeo Tsujii et al.
Neuroscience research, 67(1), 80-85 (2010-02-09)
Histamine H1 receptor antagonists (antihistamines) are widely used for the treatment of allergic disorders in young children. This study examined the effects of antihistamine on prefrontal cortex activity in preschool children using near-infrared spectroscopy (NIRS), an emerging brain-imaging method suitable
Paramdeep S Bilkhu et al.
Ophthalmology, 121(1), 72-78 (2013-09-28)
To investigate whether artificial tears and cold compress alone or in combination provide a treatment benefit and whether they were as effective as or could enhance topical antiallergic medication. Randomized, masked clinical trial. Eighteen subjects (mean age, 29.5±11.0 years) allergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service