Skip to Content
Merck
All Photos(1)

Key Documents

Y0000893

Imipramine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Imipramine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2 · HCl
CAS Number:
Molecular Weight:
316.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

imipramine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2CCc3ccccc13

InChI

1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H

InChI key

XZZXIYZZBJDEEP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Imipramine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters with Kis of 7.7 nM and 67 nM, respectively. Has little effect on the dopamine transporter (Ki = 25 μM).
Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters; has little effect on the dopamine transporter.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ritabrata Banerjee et al.
Psychiatry investigation, 11(3), 297-306 (2014-08-12)
The present study aimed to investigate whether graded doses of Bacopa Monniera (BM) extract could produce antidepressant-like effects in chronic unpredictable stress (CUS) induced depression in rats and its possible mechanism(s). Rats were subjected to an experimental setting of CUS.
Kazumi Miyawaki et al.
Journal of pharmacological sciences, 127(3), 382-390 (2015-04-04)
Biological rhythms are critical in the etiology of mood disorders; therefore, effective mood disorder treatments should address rhythm disturbances. Among the variables synchronized with the light-dark cycle, spontaneous activity in rodents is useful for investigating circadian rhythms. However, previous studies
Helena Domin et al.
Behavioural brain research, 273, 23-33 (2014-07-22)
The glutamatergic predominance in the excitatory-inhibitory balance is postulated to be involved in the pathogenesis of depression. Such imbalance may be induced by astrocyte ablation which reduces glutamate uptake and increases glutamate level in the synaptic cleft. In the present
Juliana O G Nascimento et al.
Brain research bulletin, 109, 39-45 (2014-10-08)
The dorsomedial hypothalamus (DMH) has long been associated with the regulation of escape, a panic-related defensive response. Previous evidence has shown that the activation of serotonin (5-HT) 1A and 2A receptors impairs escape behavior induced by the electrical stimulation of
Gu Ping et al.
Pharmacology, biochemistry, and behavior, 124, 5-12 (2014-05-23)
Valsartan is a synthetic non-peptide angiotensin II type 1 receptor antagonist that dilates blood vessels and reduces blood pressure by blocking the action of angiotensin, and is safe and well tolerated in hypertensive patients. Population-based studies have suggested a positive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service