Skip to Content
Merck
All Photos(1)

Key Documents

Y0000787

Caffeine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Caffeine, 1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

caffeine

manufacturer/tradename

EDQM

mp

234-236.5 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Caffeine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Bousette et al.
Journal of cellular physiology, 229(3), 374-383 (2013-09-17)
Calnexin (CNX) is an endoplasmic reticulum (ER) quality control chaperone that has been implicated in ER stress. ER stress is a prominent pathological feature of various pathologic conditions, including cardiovascular diseases. However, the role of CNX and ER stress has
Louise A Horrigan et al.
Pharmacology & therapeutics, 111(3), 877-892 (2006-03-17)
Caffeine is a member of the methylxanthine family of drugs, and is the most widely consumed behaviourally active substance in the western world. This article is focused on the impact of caffeine on immune function. In this regard, a number
Pierre Ayotte et al.
Environmental health perspectives, 113(10), 1318-1324 (2005-10-06)
The Lower North Shore region of the St. Lawrence River is home to a fish-eating population that displays an unusually high body burden of several organochlorines, including polychlorinated biphenyls (PCBs) and dioxin-like compounds (DLCs). We measured biomarkers indicative of liver
Jonathan M Hodgson et al.
The American journal of clinical nutrition, 97(5), 943-950 (2013-04-05)
Measures of blood pressure variation have been associated with cardiovascular disease and related outcomes. The regular consumption of black tea can lower blood pressure, but its effects on blood pressure variation have yet to be investigated. We aimed to assess
Markus Fehrholz et al.
American journal of physiology. Lung cellular and molecular physiology, 306(1), L101-L109 (2013-10-29)
Factors positively influencing surfactant homeostasis in general and surfactant protein B (SP-B) expression in particular are considered of clinical importance regarding an improvement of lung function in preterm infants. The objective of this study was to identify effects of physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service