Skip to Content
Merck
All Photos(1)

Key Documents

Y0000737

Flavoxate impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O4
CAS Number:
Molecular Weight:
280.27
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flavoxate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C17H12O4/c1-10-14(18)12-8-5-9-13(17(19)20)16(12)21-15(10)11-6-3-2-4-7-11/h2-9H,1H3,(H,19,20)

InChI key

KMMBBZOSQNLLMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flavoxate impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C X Zhang et al.
Journal of chromatography, 612(2), 287-294 (1993-02-26)
The effects of tetraalkylammonium salts and sodium dodecyl sulphate on the migration behaviour of human urinary components and other negatively charged or neutral solutes were investigated. The sulphate acted mainly on hydrophobic and positively charged substances, whereas the ammonium salts
M Caine et al.
Urology, 37(4), 390-394 (1991-04-01)
The antispasmodic effects of the flavone compounds flavoxate hydrochloride, 3-methylflavone carboxylic acid (MFCA), and REC 15/2053 (and in the case of the detrusor, oxybutynin), on the human detrusor, prostatic adenoma, prostatic capsule, and bladder neck, were studied by the in
P Cazzulani et al.
Arzneimittel-Forschung, 38(3), 379-382 (1988-03-01)
The pharmacological properties of 3-methylflavone-8-carboxylic acid (MFCA), the main metabolite of flavoxate, have been studied in vitro and in vivo. MFCA did not display antispasmodic activity on isolated organs contractions induced by histamine, acetylcholine or CaCl2, nor did it exhibit
Ramisetti N Rao et al.
Journal of separation science, 34(22), 3265-3271 (2011-10-15)
A novel water-compatible molecularly imprinted SPE combined with zwitterionic hydrophilic interaction liquid chromatography method for selective extraction and determination of 3-methylflavone-8-carboxylic acid (MFA), the main active metabolite of flavoxate in human urine, was developed and validated. The effects of progenic
Alaa el-Gindy et al.
Drug development and industrial pharmacy, 34(12), 1311-1322 (2008-10-14)
Liquid chromatographic method was presented for the determination of flavoxate hydrochloride (FX) and its hydrolysis product. The method was based on high-performance liquid chromatographic (HPLC) separation of FX from its hydrolysis product on CN column using a mobile phase consisting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service