Skip to Content
Merck
All Photos(1)

Key Documents

Y0000685

Letrozole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4,4′-(1H-1,2,4-Triazol-1-ylmethylene)bisbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N5
CAS Number:
Molecular Weight:
285.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

letrozole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N#CC(C=C1)=CC=C1C(N2C=NC=N2)C3=CC=C(C#N)C=C3

InChI

1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H

InChI key

HPJKCIUCZWXJDR-UHFFFAOYSA-N

Gene Information

human ... CYP19A1(1588)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Letrozole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Letrozole is a third generation nonsteroidal aromatase inhibitor. It is a competitive inhibitor of the aromatase enzyme system and thus inhibits the conversion of androgens to estrogens. Letrozole inhibits the aromatase enzyme by competitively binding to the heme of the cytochrome P450 subunit of the enzyme, resulting in a reduction of estrogen biosynthesis in all tissues.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingyi Cheng et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(3), 333-340 (2013-02-13)
Although endocrine therapy is an effective method to treat estrogen receptor (ER)-positive breast cancer, approximately 30%-40% of all hormone receptor-positive tumors display de novo resistance. The aim of our current study was to analyze whether (18)F-labeled fluoromisonidazole (1-(2-nitro-1-imidazolyl)-2-hydroxy-3-fluoropropane [(18)F-FMISO]) PET/CT
Israel Ortega et al.
Fertility and sterility, 99(3), 889-896 (2012-12-04)
To evaluate the effects of letrozole on ovarian size and steroidogenesis in vivo, as well as on proliferation and steroidogenesis of theca-interstitial cells alone and in coculture with granulosa cells using an in vitro model. In vivo and in vitro studies. Research laboratory. Immature
Mothaffar F Rimawi et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(14), 1726-1731 (2013-04-10)
We previously reported the eradication of human epidermal growth factor receptor 2 (HER2)- amplified human xenografts in mice by inhibition of the HER2 pathway with lapatinib and trastuzumab to block all homo- and heterodimer signaling as well as by blockade
Dennis C Sgroi et al.
Journal of the National Cancer Institute, 105(14), 1036-1042 (2013-07-03)
Biomarkers to optimize extended adjuvant endocrine therapy for women with estrogen receptor (ER)-positive breast cancer are limited. The HOXB13/IL17BR (H/I) biomarker predicts recurrence risk in ER-positive, lymph node-negative breast cancer patients. H/I was evaluated in MA.17 trial for prognostic performance
Rob Riemsma et al.
Current medical research and opinion, 28(8), 1263-1279 (2012-06-29)
Third-generation aromatase inhibitors (letrozole, anastrozole) have shown superior efficacy in early and advanced breast cancer compared with tamoxifen. For HR+, HER2+ MBC, combination of an AI with an anti-HER2 agent (lapatinib or trastuzumab) has shown clinical benefit. Six databases were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service