Skip to Content
Merck
All Photos(1)

Key Documents

Y0000558

Fluconazole for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fluconazole, 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC1=CC(F)=C(C(CN2N=CN=C2)(O)CN3N=CN=C3)C=C1

InChI

1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2

InChI key

RFHAOTPXVQNOHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluconazole for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Fluconazole is an antifungal agent. It is highly selective inhibitor of fungal cytochrome P-450 sterol C-14 α-demethyllation. Fluconazole is a potent inhibitor of CYP2C9. Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene.
Fluconazole is an antifungal agent; highly selctive inhibitor of fungal cytochrome P-450 sterol C-14 α-demethyllation. Potent inhibitor of CYP2C9. Interferes with fungal ergosterol synthesis; downregulates metallothionein gene.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A-C L Meyer et al.
Tropical medicine & international health : TM & IH, 18(4), 495-503 (2013-02-02)
To test the hypothesis that a screening and treatment intervention for early cryptococcal infection would improve survival among HIV-infected individuals with low CD4 cell counts. Newly enrolled patients at Family AIDS Care and Education Services (FACES) in Kenya with CD4 ≤ 100
Nicholas G Wysham et al.
Chest, 143(5), 1478-1479 (2013-05-08)
We report a case of Cryptococcus neoformans pneumonia in a patient taking ruxolitinib, a janus kinase 1,2 inhibitor approved for the treatment of myelofibrosis. We hypothesize that ruxolitinib contributed to this infection through its effects on cell-mediated immunity. Clinicians should
M C Ethier et al.
British journal of cancer, 106(10), 1626-1637 (2012-05-10)
Objectives were to compare systemic mould-active vs fluconazole prophylaxis in cancer patients receiving chemotherapy or haematopoietic stem cell transplantation (HSCT). We searched OVID MEDLINE and the Cochrane Central Register of Controlled Trials (1948-August 2011) and EMBASE (1980-August 2011). Randomised controlled
Kim C M van der Elst et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 59(11), 1527-1533 (2014-08-26)
Fluconazole is recommended as first-line treatment in invasive candidiasis in children and infants. Although timely achievement of adequate exposure of fluconazole improves outcome, therapeutic drug monitoring is currently not recommended. We conducted a retrospective study of critically ill children treated
Jana Bogum et al.
Journal of the American Society of Nephrology : JASN, 24(5), 744-758 (2013-04-06)
In the principal cells of the renal collecting duct, arginine vasopressin (AVP) stimulates the synthesis of cAMP, leading to signaling events that culminate in the phosphorylation of aquaporin-2 water channels and their redistribution from intracellular domains to the plasma membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service