Skip to Content
Merck
All Photos(1)

Key Documents

Y0000541

Thioridazine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Thioridazine, 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylthio)-10H-phenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2S2
CAS Number:
Molecular Weight:
370.57
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thioridazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C21H26N2S2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(24-2)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3

InChI key

KLBQZWRITKRQQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thioridazine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

It is anticipated that as is usually the case, financial motivation will move TZ from its current status “potential anti-MDR/XDR agent,” to one that will cure MDR and XDR TB. Foreword.
Leonard Amaral
Recent patents on anti-infective drug discovery, 6(2), 76-76 (2011-07-30)
Shen-Chieh Chang et al.
Therapeutic drug monitoring, 34(3), 345-348 (2012-05-10)
In this study, the authors studied the effect of thioridazine (TDZ) on the pharmacokinetic profile of quetiapine (QTP) in Taiwanese patients with schizophrenia. Sixteen subjects with schizophrenia were recruited for this study. The authors pretreated 8 patients with TDZ 50
Jørn B Christensen et al.
PloS one, 8(3), e57493-e57493 (2013-03-19)
A long list of chemotherapeutical drugs used in the treatment of the peripheral and the central nervous systems possess anti-microbial activity. Some of these neurotropic compounds are chiral, with the one stereo isomeric form exaggerating reduced neurotropism. This is the
Diana Machado et al.
PloS one, 7(4), e34538-e34538 (2012-04-12)
Multidrug resistant (MDR) tuberculosis is caused by Mycobacterium tuberculosis resistant to isoniazid and rifampicin, the two most effective drugs used in tuberculosis therapy. Here, we investigated the mechanism by which resistance towards isoniazid develops and how overexpression of efflux pumps
Eduardo Abbate et al.
The Journal of antimicrobial chemotherapy, 67(2), 473-477 (2011-12-03)
Current drug choices to treat extensively drug-resistant (XDR) tuberculosis (TB) are scarce; therefore, information on the safety, tolerability and efficacy of alternative regimens is of utmost importance. The aim of this study was to describe the management, drug adverse effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service