Skip to Content
Merck
All Photos(1)

Key Documents

Y0000438

Coumarin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Beilstein:
383644
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor pressure

0.01 mmHg ( 47 °C)

API family

coumarin

manufacturer/tradename

EDQM

bp

298 °C (lit.)

mp

68-73 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1Oc2ccccc2C=C1

InChI

1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Coumarin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donglei Yu et al.
Medicinal research reviews, 23(3), 322-345 (2003-03-21)
Numerous plant-derived compounds have been evaluated for inhibitory effects against HIV replication, and some coumarins have been found to inhibit different stages in the HIV replication cycle. This review article describes recent progress in the discovery, structure modification, and structure-activity
Musiliyu A Musa et al.
Current medicinal chemistry, 15(26), 2664-2679 (2008-11-11)
The coumarin (benzopyran-2-one, or chromen-2-one) ring system, present in natural products (such as the anticoagulant warfarin) that display interesting pharmacological properties, has intrigued chemists and medicinal chemists for decades to explore the natural coumarins or synthetic analogs for their applicability
Pravin O Patil et al.
Bioorganic & medicinal chemistry, 21(9), 2434-2450 (2013-03-23)
Monoamine oxidase (MAO) enzyme inhibition is a crucial target for the management of depression and Alzheimer disease and inhibitors of MAO are the most important drugs for their management. Coumarins are a large family of compounds, of natural and synthetic
Klaus Abraham et al.
Molecular nutrition & food research, 54(2), 228-239 (2009-12-22)
Coumarin is a secondary phytochemical with hepatotoxic and carcinogenic properties. For the carcinogenic effect, a genotoxic mechanism was considered possible, but was discounted by the European Food Safety Authority in 2004 based on new evidence. This allowed the derivation of
B G Lake
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 37(4), 423-453 (1999-07-27)
The metabolism, toxicity and results of tests for carcinogenicity have been reviewed with respect to the safety for humans of coumarin present in foodstuffs and from fragrance use in cosmetic products. Coumarin is a natural product which exhibits marked species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service