Skip to Content
Merck
All Photos(1)

Key Documents

Y0000009

Fluphenazine enantate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[4-[3-[2-(Trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazinyl]ethyl heptanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38F3N3O2S
CAS Number:
Molecular Weight:
549.69
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluphenazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc2c(cc1)Sc3c(cccc3)N2CCCN4CCN(CC4)CCOC(=O)CCCCCC

InChI

1S/C29H38F3N3O2S/c1-2-3-4-5-11-28(36)37-21-20-34-18-16-33(17-19-34)14-8-15-35-24-9-6-7-10-26(24)38-27-13-12-23(22-25(27)35)29(30,31)32/h6-7,9-10,12-13,22H,2-5,8,11,14-21H2,1H3

InChI key

LRWSFOSWNAQHHW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluphenazine enantate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Glazer
The Journal of clinical psychiatry, 45(5 Pt 2), 28-35 (1984-05-01)
The risks and benefits associated with depot fluphenazine are reassessed by a review and critique of the literature, with an emphasis on controlled studies comparing depot to oral preparations. Specific gaps in our knowledge are noted and recommendations are made
K Yamada et al.
Pharmacology, 24(3), 147-155 (1982-01-01)
The cataleptic state induced by the depot-type preparations of fluphenazine (FZ) was investigated in mice. Intramuscular injections of FZ-enanthate or FZ-decanoate produced a marked and long-lasting catalepsy, which was antagonized by trihexyphenidyl, biperiden and promethazine but not by l-dopa. These
J P Lanza et al.
Comptes rendus des seances de la Societe de biologie et de ses filiales, 173(4), 797-806 (1979-01-01)
The effects of two neuroleptics (pipotiazine and fluphenazine) and five long-acting neuroleptics (pipotiazine undecylenate and palmitate, fluphenazine enanthate and decanoate, fluopentixol decanoate) are tested in the rat, during an observation period of 20 to 40 days following only one injection
A C Altamura et al.
Psychopharmacology, 87(1), 30-33 (1985-01-01)
Seven outpatients already receiving neuroleptic drugs by depot intramuscular injections were treated in two consecutive 3-week periods with 25 mg fluphenazine doses as enanthate and decanoate esters in a double-blind crossover study. They were assessed for incidence of akinesia, involuntary
Fluphenazine enanthate and fluphenazine decanoate in the treatment of schizophrenic outpatients: extrapyramidal symptoms and therapeutic effect.
G Chouinard et al.
The American journal of psychiatry, 139(3), 312-318 (1982-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service