VT000903
Escherichia coli WDCM 00090 Vitroids™
80 - 130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
E. coli WDCM 00090 Vitroids™
About This Item
Recommended Products
grade
certified reference material
Quality Level
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
microbiology
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Download your certificate at: http://www.sigma-aldrich.com.
Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service