Skip to Content
Merck

V5262

Veal infusion broth

suitable for microbiology, NutriSelect® Plus

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41171607
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

clinical testing
food and beverages
life science and biopharma, microbiology

suitability

nonselective for Neisseria spp., nonselective for Staphylococcus spp., nonselective for Streptococcus spp.

Application

Recommended for the cultivation of fastidious pathogenic bacteria.

Preparation Note

Suspend 25 g of Veal Infusion Broth in 1000 ml of distilled water. Boil to dissolve the medium completely. Dispense and sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes.

Other Notes

Ingredients
Veal infusion (from 500 g) 10g/L
Proteose peptone 10 g/L
Sodium chloride 5g/L
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library