Skip to Content
Merck
All Photos(1)

Key Documents

T6750

Sigma-Aldrich

Thioglycolic acid solution

~70 % (w/w) in H2O

Synonym(s):

Mercaptoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOH
CAS Number:
Molecular Weight:
92.12
Beilstein:
506166
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

concentration

~70 % (w/w) in H2O

density

1.25 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)CS

InChI

1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)

InChI key

CWERGRDVMFNCDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

At room temperature, concentrations over about 70% in water tend to form 1-2% thioglycolides per month which hydrolyze to the original free compound when made acid or alkaline. The 70% solution oxidizes in air but is stable at room temperature when tightly closed. Thioglycolate salts may also lose purity on storage. The exclusion of air does not materially improve stability.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baofu Han et al.
Colloids and surfaces. B, Biointerfaces, 100, 209-214 (2012-07-07)
Fluorescent carbon dots (CDs) were solvothermaly synthesized in water-glycol medium by using glucose as carbon source and then modified with polyethyleneimine (PEI) for the first time to improve fluorescence quality. The as-prepared CDs were monodispersed sphere particles with a diameter
Jun-Ichi Wachino et al.
Antimicrobial agents and chemotherapy, 57(1), 101-109 (2012-10-17)
A novel subclass B3 metallo-β-lactamase (MBL), SMB-1, recently identified from a Serratia marcescens clinical isolate, showed a higher hydrolytic activity against a wide range of β-lactams than did the other subclass B3 MBLs, i.e., BJP-1 and FEZ-1, from environmental bacteria.
Markus Aswendt et al.
Nanomedicine (London, England), 10(16), 2499-2512 (2015-08-22)
We validated novel bimodal iron oxide particles as substitute of ferumoxides for efficient labeling of human neural stem cells (NSCs). The dextrane-coated FeraTrack Direct (FTD)-Vio particles have additional far-red fluorophores for microscopic cell analysis. MR relaxometry, spectrophotometric iron determination and
Zhengqing Liu et al.
Analytica chimica acta, 745, 78-84 (2012-09-04)
A novel fluorescent probe for Cu(2+) determination based on the fluorescence quenching of glyphosate (Glyp)-functionalized quantum dots (QDs) was firstly reported. Glyp had been used to modify the surface of QDs to form Glyp-functionalized QDs following the capping of thioglycolic
Gizem Ayna et al.
PloS one, 7(6), e40069-e40069 (2012-07-07)
Pathogen-activated and damage-associated molecular patterns activate the inflammasome in macrophages. We report that mouse macrophages release IL-1β while co-incubated with pro-B (Ba/F3) cells dying, as a result of IL-3 withdrawal, by apoptosis with autophagy, but not when they are co-incubated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service