T59307
Triethyl borate
99%
Synonym(s):
Boric acid triethyl ester, Boron ethoxide, Triethoxyborane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5O)3B
CAS Number:
Molecular Weight:
145.99
Beilstein:
1699052
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
5.04 (vs air)
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.374 (lit.)
bp
117-118 °C (lit.)
density
0.858 g/mL at 25 °C (lit.)
SMILES string
CCOB(OCC)OCC
InChI
1S/C6H15BO3/c1-4-8-7(9-5-2)10-6-3/h4-6H2,1-3H3
InChI key
AJSTXXYNEIHPMD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Triethyl borate can be used as boron source to synthesize lithium borophosphates, alumina borosilicate ceramic nanofibres and boron-doped single wall carbon nanotubes.[74}
Triethyl borate may be used as boron source to synthesize lithium borophosphates, alumina borosilicate ceramic nanofibres and boron-doped single wall carbon nanotubes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sol-gel lithium borophosphates
Ali A F
J. Mater. Sci., 14(4), 1510-1515 (1999)
Electrospinning and characterization of alumina borosilicate ceramic nanofibres.
Tanriverdi S, et al.
Materials Science-Poland, 25(4), 957-968 (2007)
Sol-gel lithium borophosphates.
Ali AF, et al.
Journal of Materials Research, 14(4), 1510-1515 (1999)
Production and characterization of boron-doped single wall carbon nanotubes.
Monteiro F H
The Journal of Physical Chemistry, 116(5), 3281-3285 (2012)
Production and characterization of boron-doped single wall carbon nanotubes.
Monteiro FH, et al.
The Journal of Physical Chemistry C, 116(5), 3281-3285 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service