Skip to Content
Merck
All Photos(1)

Documents

T1500000

Tobramycin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Nebramycin Factor 6, O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1→4)]-2-deoxy-D-streptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37N5O9
CAS Number:
Molecular Weight:
467.51
Beilstein:
1357507
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

API family

tobramycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@@H](N)[C@H]3O)[C@H]2O)[C@H](N)C[C@@H]1O

InChI

1S/C18H37N5O9/c19-3-9-8(25)2-7(22)17(29-9)31-15-5(20)1-6(21)16(14(15)28)32-18-13(27)11(23)12(26)10(4-24)30-18/h5-18,24-28H,1-4,19-23H2/t5-,6+,7+,8-,9+,10+,11+,12+,13+,14-,15+,16-,17+,18+/m0/s1

InChI key

NLVFBUXFDBBNBW-SNGYORCQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tobramycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Tobramycin is an aminoglycoside.
Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding.
Spectrum of Activity: Gram negative bacteria. Not effective against Enterococci.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blanca Rojas et al.
Journal of neuroinflammation, 11, 133-133 (2014-07-30)
Glaucomatous optic neuropathy, a leading cause of blindness, can progress despite control of intraocular pressure - currently the main risk factor and target for treatment. Glaucoma progression shares mechanisms with neurodegenerative disease, including microglia activation. In the present model of
Susan M Cheer et al.
Drugs, 63(22), 2501-2520 (2003-11-12)
Specifically formulated for nebulisation using the PARI LC PLUS reusable nebuliser, tobramycin solution for inhalation (TSI) [TOBI] provides a high dose of tobramycin (an aminoglycoside antibacterial with good activity against Pseudomonas aeruginosa) to the lungs of patients with cystic fibrosis
David E Geller et al.
Chest, 122(1), 219-226 (2002-07-13)
To describe the pharmacokinetics and bioavailability of inhaled tobramycin (TOBI; Chiron Corporation; Seattle, WA), 300-mg dose, delivered by a nebulizer (PARI LC Plus; Pari Respiratory; Richmond, VA) and a compressor (Pulmo-Aide, model 5650D; DeVilbiss Health Care; Somerset, PA) in cystic
David E Geller et al.
Journal of aerosol medicine and pulmonary drug delivery, 24(4), 175-182 (2011-03-15)
Abstract At present, the only approved inhaled antipseudomonal antibiotics for chronic pulmonary infections in patients with cystic fibrosis (CF) are nebulized solutions. However, prolonged administration and cleaning times, high administration frequency, and cumbersome delivery technologies with nebulizers add to the
Yu-Chi Liu et al.
Investigative ophthalmology & visual science, 55(10), 6736-6741 (2014-09-25)
To correlate the degree of anterior chamber (AC) inflammation and corneal thickness evaluated by anterior segment optical coherence tomography (ASOCT) with corneal graft rejection status and to explore the value of ASOCT in assisting the diagnosis or prediction of graft

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service