Skip to Content
Merck

SYNPLE-P005

Reagent-cartridge CRBN-PEG5

Synple

Synonym(s):

Synple 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


ligand

pomalidomide

Quality Segment

reaction suitability

reaction type: PROTAC formation

parameter

up to 0.1 mmol scale

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

General description

Reagent-cartridge CRBN-PEG5 is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the synthesis of molecules for targeted protein degradation and PROTAC® (proteolysis-targeting chimeras) research. The cartridge contains all necessary reagents for linking the user selected aldehyde or ketone to the amine terminated linker through a reductive amination. This cartridge features a Cereblon (CRBN) recruiting ligand (pomalidomide) and PEG linker. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation.

Scale: up to 0.1 mmol
Linker: PEG5
E3 ligase target and ligand: Cereblon (CRBN), pomalidomide
Terminal functional group: amine
Compatible substrates: ketone, aldehyde

Cartridge contents:
  • Reducing agent (supported cyanoborohydride)
  • Base (supported triethylamine)
  • SCX (for purification)
  • Degrader building block linker reagent

Related Cartridges:
  • SYNPLE-P001Reagent-cartridge CRBN-PEG1
  • SYNPLE-P002Reagent-cartridge CRBN-PEG2
  • SYNPLE-P003Reagent-cartridge CRBN-PEG3
  • SYNPLE-P004Reagent-cartridge CRBN-PEG4
  • SYNPLE-P021Reagent-cartridge VHL-PEG1
  • SYNPLE-P022Reagent-cartridge VHL-PEG2
  • SYNPLE-P023Reagent-cartridge VHL-PEG3
  • SYNPLE-P024Reagent-cartridge VHL-PEG4
  • SYNPLE-P025Reagent-cartridge VHL-PEG5

Learn more about the Synple automated synthesis platform.

Learn more about protein degradation.

Explore all our Protein Degrader Building Blocks

Features and Benefits of Synple Platform
  • Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
  • Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety - all chemicals are contained in the system, minimizing the user′s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system - simple installation
  • Reaction temperatures from 25 °C - 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library