reaction suitability
reaction type: Biotinylations
parameter
up to 0.1 mmol scale
SMILES string
[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C
InChI
1S/C28H38N4O7S/c1-17-24(40-16-30-17)19-7-5-18(6-8-19)14-29-26(37)21-13-20(33)15-32(21)27(38)25(28(2,3)4)31-22(34)9-11-39-12-10-23(35)36/h5-8,16,20-21,25,33H,9-15H2,1-4H3,(H,29,37)(H,31,34)(H,35,36)/t20-,21+,25-/m1/s1
InChI key
HVPZHRHEHKZOOJ-TYBLODHISA-N
General description
Reagent-cartridge Biotin tag-PEG3-COOTfp is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the biotin tagging of amines through an acylation reaction. The cartridge contains all necessary reagents for linking the chosen amine substrate to biotin.
Scale: up to 0.1 mmol
Linker: PEG3
Terminal functional group: active ester
Compatible substrates: amine, amine salts
Cartridge contents:
Related Cartridges:
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
Scale: up to 0.1 mmol
Linker: PEG3
Terminal functional group: active ester
Compatible substrates: amine, amine salts
Cartridge contents:
- Base (supported triethylamine)
- Silica supported carbonate (for purification)
- Biotin linker
Related Cartridges:
- SYNPLE-BT002Reagent-cartridge Biotin tag-PEG2-NH2
- SYNPLE-BT003Reagent-cartridge Biotin tag-PEG3-NH2
- SYNPLE-BT004Reagent-cartridge Biotin tag-PEG4-NH2
- SYNPLE-BT012Reagent-cartridge Biotin tag-PEG2-O-4-P-NH
- SYNPLE-BT013Reagent-cartridge Biotin tag-PEG3-O-4-P-NH
- SYNPLE-BT014Reagent-cartridge Biotin tag-PEG4-O-4-P-NH
- SYNPLE-BT022Reagent-cartridge Biotin tag-PEG2-a-COOTfp
- SYNPLE-BT023Reagent-cartridge Biotin tag-PEG3-a-COOTfp
- SYNPLE-BT024Reagent-cartridge Biotin tag-PEG4-a-COOTfp
- SYNPLE-BT032Reagent-cartridge Biotin tag-PEG2-COOTfp
- SYNPLE-BT034Reagent-cartridge Biotin tag-PEG4-COOTfp
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
- Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
- Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety - all chemicals are contained in the system, minimizing the user′s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system - simple installation
- Reaction temperatures from 25 °C - 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Legal Information
Product of Synple Chem AG
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
