Skip to Content
Merck

STS0200

ECO TWEEN® 20

greener alternative

viscous liquid

Synonym(s):

TWEEN® 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

assay

≥40%

form

viscous liquid

mol wt

~1228

composition

lauric acid, ≥40% (balance primarily myristic, palmitic, and stearic acids)

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

≥40.0% (GC)

impurities

≤3.0% water

refractive index

n20/D 1.468 (lit.)

CMC

0.06 mM (20-25°C)

mp

98.9 °C

acid number

≤2.2 mg KOH/g

hydroxyl value

96‑108 mg KOH/g

transition temp

cloud point 76 °C

solubility

water: 100 mg/mL, clear, faintly yellow to yellow

density

1.095 g/mL at 25 °C (lit.)

HLB

16.7

greener alternative category

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

ECO TWEEN® 20, also known as Polyoxyethylenesorbitan monolaurate, is a bio-based, nonionic detergent manufactured from a natural fatty acid. This surfactant, derived from natural sources, provides various functional advantages, such as detergency, emulsification, and wetting. Its versatility makes it well-suited for a variety of applications in biochemical and biological research, with common uses including cell lysis, protein extraction, nuclei isolation and cell fractionation.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

TWEEN 20 is a polyoxyethylene sorbitol ester that belongs to the polysorbate family. It is a nonionic detergent having a molecular weight of 1,225 daltons, assuming 20 ethylene oxide units, 1 sorbitol, and 1 lauric acid as the primary fatty acid. The ethylene oxide subunits are responsible for the hydrophilic nature of the surfactant, while the hydrocarbon chains provide the hydrophobic environment. Sorbitol forms the backbone ring to which the ethylene oxide polymers are attached.

Application

Non-ionic detergent
TWEEN 20 is a nonionic detergent widely used in biochemical applications. It has been used as an emulsifying agent for the preparation of stable oil-in-water emulsions. TWEEN 20 has been used in pre-extraction of membranes to remove peripheral proteins (used at 2% for extraction of membrane-bound proteins).
TWEEN 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%. TWEEN 20 can be used for lysing mammalian cells at a concentration of 0.005 to 0.5%.
TWEEN 20 has been used as a component of:
  • washing buffer and blocking buffer in Western blotting
  • blocking buffer in immunohistochemistry
  • washing buffer in ELISA (enzyme linked immunosorbent assay)
  • reaction mixture in PCR (polymerase chain reaction)

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Preparation Note

TWEEN 20 is miscible in water (100mg/mL), yielding a clear, yellow solution. It is also miscible with alcohol, dioxane, and ethyl acetate. TWEEN 20 is insoluble in liquid paraffin and fixed oils.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

ECO TWEEN is a registered trademark of Croda Inc.
TWEEN is a registered trademark of Croda International PLC


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

527.0 °F - Pensky-Martens closed cup

flash_point_c

275 °C - Pensky-Martens closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Gene expression and immune response kinetics using electroporation-mediated DNA delivery to muscle.
Gr?nevik E et al
The journal of gene medicine, 7(2), 218-227 (2005)
Use of tRNA consensus primers to indicate subgroups of Pseudomonas solanacearum by polymerase chain reaction amplification.
Seal SE et al
Applied and Environmental Microbiology, 58(11), 3759-3761 (1992)
Polysorbates 20 and 80 used in the formulation of protein biotherapeutics: structure and degradation pathways.
Kerwin BA
J. Pharm. Pharm. Sci., 97(8), 2924-2935 (2008)



Global Trade Item Number

SKUGTIN
STS0200-250ML04065266006629
STS0200-1L04065266006612